missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fluo-5F, Pentapotassium Salt, cell impermeant
Description
Analog of fluo-4 with lower Ca2+-binding affinity, making it suitable for detecting intracellular calcium levels in 1μM to 1 mM range that would saturate esponse of fluo-3 and fluo-4
Labeled calcium indicators are molecules that exhibit an increase in fluorescence upon binding Ca2+.
- Cells may be physically loaded with cell-impermeant salt forms of these indicators using patch pipet or microinjection or Influx™ pinocytotic cell-loading reagent
- Compatible with excitation at 488nm by argon-ion laser sources, making them useful for confocal microscopy, flow cytometry, and microplate screening applications
- Exhibit fluorescence increase upon binding Ca2+ with little shift in wavelength
- BAPTA-based indicators bind various heavy metal cations (e.g., Mn2+, Zn2+, Pb2+) with substantially higher affinity than Ca2+
- Perturbations to calcium measurements caused by presence of the ions can be controlled using heavy metal-selective chelator
- Label (Ex/Em of Ca2+-bound form): Fluo-5N (494/516nm)
- Fluorescence intensity increase upon binding Ca2+: >100 fold
- Kd for Ca2+ in buffer: approximately 90μM
Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Content And Storage | Store in freezer -5°C to -30°C and protect from light. |
| Detection Method | Fluorescence |
| For Use With (Equipment) | Confocal Microscope, Fluorescence Microscope, High Content Analysis Instrument, HTS Reader, Microplate Reader, Fluorescent Imager |
| Product Type | Calcium Indicator |
| Dye Type | Fluorescent Dye-Based |
| Quantity | 500 μg |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.