missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Filmtracer™ LIVE/DEAD™ Biofilm Viability Kit

Catalog No. L10316
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L10316 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L10316 Supplier Invitrogen™ Supplier No. L10316

Includes

2 ingredients

Uses mixture of green fluorescent and red fluorescent stains

The FilmTracer LIVE/DEAD Biofilm Viability kit provides a two-color fluorescence assay of bacterial viability, based on membrane integrity, that has proven useful for a diverse array of bacterial genera including those growing in biofilm communities. The LIVE/DEAD Biofilm Viability kit utilizes mixtures of SYTO™ 9 green fluorescent nucleic acid stain and the red-fluorescent nucleic acid stain, propidium iodide. These stains differ both in their spectral characteristics and in their ability to penetrate healthy bacterial cells. When used alone, the SYTO 9 stain generally labels all bacteria in a population: those with intact membranes and those with damaged membranes. In contrast, propidium iodide penetrates only bacteria with damaged membranes, causing a reduction in the SYTO 9 stain fluorescence with both dyes are present. Thus, with an appropriate mixture of the SYTO 9 and propdium iodide stains, bacteria with intact cell membranes stain fluorescent green, whereas bacteria with damaged membranes stain fluorescent red.

Analysis of Bacteria, Bacterial Viability and Cytotoxicity Analysis, Cell Analysis, Microbiological Analysis

Order Info

Shipping Condition: Room temperature

Specifications

Color Red, Green
Content And Storage 2 ingredients, store in freezer -5°C to -30°C; protect from light
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Confocal Microscope, Fluorescence Microscope, Flow Cytometer, Microplate Reader, Fluorescent Imager
Product Type Biofilm Viability Kit
Dye Type SYTO™ 9, propidium iodide
Emission Visible
Form Liquid
Product Line FilmTracer, Molecular Probes
Quantity 1 kit
Shipping Condition Room Temperature
Sub Cellular Localization Cytoplasm & Cytosol
Show More Show Less
How do I prepare dead cell controls for LIVE/DEAD cell viability assays?

There are two easy options. One is to heat-inactivate the cells by placing at 60 degrees C for 20 minutes. The second is to subject the cells to 70% ethanol. Alcohol-fixed cells can be stored indefinitely in the freezer until use, potentially up to several years.

Centrifuge cells, pellet, and remove supernatant.
Fix cells: Add 10 mL ice cold 70% ETOH to a 15 mL tube containing the cell pellet, adding dropwise at first while vortexing, mix well.
Store in freezer until use.
When ready to use, wash twice and resuspend in buffer of choice.

For Research Use Only. Not for use in diagnostic procedures.