missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fibrinogen From Human Plasma, Alexa Fluor™ 647 Conjugate

Catalog No. f35200
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F35200 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F35200 Supplier Invitrogen™ Supplier No. F35200

For investigating platelet activation and subsequent fibrinogen binding

Molecular Probes fibrinogen conjugates are prepared by attaching fluorescent dye molecules to purified human fibrinogen (approximately 15 dye molecules for each fibrinogen molecule), purifying the conjugate to remove unreacted dye, and then lyophilizing for storage. Fluorescently labeled fibrinogen has proven to be a valuable tool for investigating platelet activation and subsequent fibrinogen binding. For instance, fluorescein-labeled fibrinogen has been used to detect fibrinogen bound to activated platelets by flow cytometry.

  • Label (Ex/Em): Alexa Fluor 647 (approx. 650/668nm)
  • Spectrally similar to Cy™5 dye conjugates
  • Lyophilized product can be dissolved in buffer (e.g., sodium bicarbonate, pH 8.3) for use
  • Fluorescence is typically detected using fluorescence microscopy or flow cytometry

Cell Adhesion, Cell Analysis, Cell Viability, Proliferation and Function, Cellular Imaging, Endocytosis, Exocytosis and Phagocytosis, Immunofluorescence (IF), Immunofluorescence Staining and Detection, In Vivo Imaging

Order Info

Shipping Condition: Room temperature

Specifications

Quantity 5 mg
Conjugate Alexa Fluor 647
Species Human
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label Type Alexa Fluor Dyes
Shipping Condition Room Temperature
Form Lyophilized
Protein Subtype Fibrinogen
Product Line Alexa Fluor

For Research Use Only. Not for use in diagnostic procedures.