missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ER-Tracker™ Blue-White DPX, for live-cell imaging

Catalog No. e12353
Encompass
Change view
Click to view available options
Quantity:
20 x 50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12353 20 x 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E12353 Supplier Invitrogen™ Supplier No. E12353

For live-cell imaging

ER-Tracker Blue-White DPX dye is a photostable probe that is selective for the endoplasmic reticulum (ER) in live cells. The cells can be aldehyde-fixed after staining with the dye but significant fluorescence signal will be lost. With excitation at ∽374nm, the environmentally-sensitive probe exhibits emission between 430 and 640nm. A long-pass DAPI filter works best.

Cell Analysis, Cell Structure, Cell Tracing & Tracking, Cellular Imaging, Endoplasmic Reticulum (ER), Immunofluorescence (IF), Immunofluorescence Staining & Detection, Organelle Tracing

Order Info

Shipping Condition: Room Temperature

Specifications

Color Blue
Concentration 1 mM
Content And Storage Store in freezer -5°C to -30°C and protect from light.
For Use With (Equipment) Fluorescence Microscope
Quantity 20 x 50 μL
Detection Method Fluorescence
Product Line ER-Tracker
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Dye
Sub Cellular Localization Endoplasmic Reticulum
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.