Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Epidermal Growth Factor, Tetramethylrhodamine Conjugate (rhodamine EGF) DFS Item

Constructed with the dye directly attached to EGF

Supplier:  Invitrogen™ E3481

Catalog No. E3481


Only null left
Explore available promotions

Description

Description

Fluorescently labeled EGF has enabled scientists to investigate receptor-membrane interactions, study receptor distribution, calculate rate constants for the interaction of EGF with its receptor, and more.

  • EGF molecule: 53 amino acids, MW=6,045 Da
  • Number of fluorophore molecules on each EGF molecule: 1
  • Fluorescence is typically monitored using a flow cytometer, fluorescence microscope, or fluorimeter

Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Wet Ice

Specifications

Specifications

Store in freezer (-5 to -30°C) and protect from light.
Fluorescence
Growth Factor
20 μg
Mammalian Cells
Solid
Classic Dyes
Wet Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.