missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Epidermal Growth Factor, Biotinylated, complexed to Alexa Fluor™ 488 Streptavidin (Alexa Fluor™ 488 EGF complex)

Catalog No. E13345
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E13345 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E13345 Supplier Invitrogen™ Supplier No. E13345

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The biotinylated conjugates utilize biotin-XX, which contains a long spacer arm designed to enhance the probe affinity for the EGF receptor

Fluorescently labeled EGF has enabled scientists to investigate receptor-membrane interactions, study receptor distribution, calculate rate constants for the interaction of EGF with its receptor, and more.

  • Cconjugates incorporating fluorescein (FITC), Oregon Green™ 514, and tetramethylrhodamine were constructed with dye directly attached to EGF
  • EGF molecule: 53 amino acids, MW=6,045 Da
  • Label (Ex/Em): Biotin-XX complexed with Alexa Fluor 488 streptavidin (approx. 495/519nm)
  • Number of fluorophore molecules on each EGF molecule: approximately 2 to 3
  • Fluorescence is typically monitored using a flow cytometer, fluorescence microscope, or fluorimeter

Bioproduction, Cell Culture, Mammalian Cell Culture, Primary Cell Culture, T Cell Culture

Order Info

Shipping Condition: Wet Ice

Specifications

Cell Type Mammalian Cell
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Detection Method Fluorescence
Product Type Growth Factor
Dye Type Alexa Fluor Dyes
Form Solid
Product Line Alexa Fluor
Quantity 100 μg
Shipping Condition Wet Ice
Protein Family EGF
What is the molecular weight of Epidermal Growth Factor, Biotinylated, complexed to Alexa Fluor 488 Streptavidin (Alexa Fluor 488 EGF complex) (Cat. No. E13345)?

The Epidermal Growth Factor, Biotinylated, complexed to Alexa Fluor 488 Streptavidin (Alexa Fluor 488 EGF complex) (Cat. No. E13345) has an approximate MW of ~60 kDa.

For Research Use Only. Not for use in diagnostic procedures.