missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EnzChek™ Reverse Transcriptase Assay Kit DFS Item

The EnzChek Reverse Transcriptase Assay Kit is a convenient, efficient, and inexpensive assay for measuring reverse transcriptase activity and screening reverse transcriptase inhibitors.

Supplier:  Invitrogen™ E22064

Numéro de catalogue. E22064

Il en reste null
Découvrez plus d'offres spéciales

Description

Description

The EnzChek Reverse Transcriptase Assay Kit is a convenient, efficient, and inexpensive assay for measuring reverse transcriptase activity and screening reverse transcriptase inhibitors. In less than an hour, samples can be read in a fluorometer or microplate reader with filter sets appropriate for fluorescein (FITC), GFP, or Alexa Fluor 488.

Features of the EnzChek Reverse Transcriptase Assay Kit include:
• Detect as little as 0.02 units of HIV-1 reverse transcriptase
• Large dynamic assay range, detect up to a 50-fold linear range
• Simple to use assay, amenable for automated high-throughput screening applications

The EnzChek Reverse Transcriptase Assay Kit uses the PicoGreen reagent, which preferentially detects dsDNA or RNA-DNA heteroduplexes over single-stranded nucleic acids or free nucleotides. In this assay, the reverse transcriptase activity in a biological sample generates long RNA-DNA heteroduplexes from a mixture of a long poly(A) template, an oligo-dT primer, and dTTP. The RNA-DNA heteroduplexes formed are then detected by the PicoGreen reagent.

Reverse transcriptase is an enzyme that functions as an RNA-dependent DNA-polymerase to synthesize DNA from an RNA template. Retroviruses such as HIV use reverse transcriptase activity to make a DNA copy of their viral RNA to insert into a host cell genome. Reverse transcriptase enzymes are also used in various cell functions, such as the enzyme telomerase, which maintains telomere length and chromosome stability and has a role in cancer and aging. In molecular biology, reverse transcriptase function is utilized to generate complementary DNA (cDNA) libraries from mRNA and quantify mRNA expression through amplification in RT-PCR.

The EnzChek Reverse Transcriptase Assay Kit provides a simple method to determine reverse transcriptase activity that is useful in the study of retroviruses and screening reverse transcriptase inhibitors, which can function as therapeutic drugs to target retroviral diseases. Because this method is faster and less expensive than standard isotopic assays or immunoassays, it is suitable for testing large numbers of biological samples, enabling automated high-throughput screening of reverse transcriptase inhibitors.

Order Info

Shipping Condition: Room temperature

Spécifications

Spécifications

Upon receipt, the kit should be stored at –20°C and protected from light. The 20X TE buffer and the lambda DNA standard are best stored at 4°C; however, either may be frozen for long term storage. Stored properly, the kit's components should remain stable for at least six months. Allow reagents to warm to room temperature before opening the vials.

Reverse Transcriptase Assay
EnzChek Assay Kit
1000 Assays
1000 Assays
Fluorescence Intensity
Fluorescence Microplate Reader
EnzChek
Room Temperature
Reverse Transcriptase
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.