Learn More
Invitrogen™ EnzChek™ Protease Assay Kit, green fluorescence, 100-1000 assays
Description
Continuously measure protease kinetics
The EnzChek™ Protease Assay Kit, green fluorescence, is a fast, simple, and direct fluorescence-based assay for detecting metallo-, serine, acid, and sulfhydryl proteases. The accompanying increase in fluorescence, which can be measured with fluorescence microplate reader, is proportional to protease activity.
- Generalized mechanism allows for measurement of a variety of proteases
- Can be used to continuously measure protease kinetics
- Excitation/emission of 505/513 nm allows use of standard fluorescein (FITC) filter settings
This EnzChek™ Protease Assay Kit contains a casein derivative that has been extensively labeled with the pH-insensitive, green-fluorescent BODIPY™ FL dye, which results in a quenching of the fluorescent dye. Protease-catalyzed hydrolysis releases the highly-fluorescent BODIPY™ FL dye-labeled peptides, allowing for quantitative detection of protease activity in solution. The green-fluorescent BODIPY™ FL dye has excitation and emission spectra similar to those of fluorescein.
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Content And Storage | Store in freezer -5°C to -30°C and protect from light. |
| Detection Method | Fluorescence |
| For Use With (Application) | Enzyme and Metabolic Activity |
| For Use With (Equipment) | Microplate Reader |
| Product Type | EnzChek Assay Kit |
| Excitation Wavelength Range | 495 to 510 nm |
| Product Line | EnzChek |
| Quantity | 1 Kit |
| Research Category | Differentiation |
| Shipping Condition | Room Temperature |
| Show More |
Frequently Asked Questions (FAQs)
The digestion buffer provided in the kit (pH 7.8) is recommended for detecting the protease activity of most proteolytic enzymes with activity optima from pH 7.4 to 8.0. However, if you are working with an enzyme that requires activation compounds or a unique pH environment, then prepare the specific buffer required in place of the digestion buffer.
As stated in the manual (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/mp06638.pdf) on page 2 under section "Storage and Handling", these kits are stable for 6-12 months if stored as indicated.
For Research Use Only. Not for use in diagnostic procedures.