missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EnzChek™ Phospholipase A1 Assay Kit

Catalog No. E10219
Encompass
Change view
Click to view available options
Quantity:
200 Reactions kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10219 200 Reactions kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E10219 Supplier Invitrogen™ Supplier No. E10219

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

1 kit; Sufficient material is supplied for 200 reactions in 96-well microplates at 100 μL per well as described in the protocol below or 400 reaction in 384 low volume microplates at a volume of ≤25 μL per well.

For continuous monitoring of phospholipase A1 (PLA1) in purified enzyme preparations, cell lysates and living cells

This kit takes the stand-alone assay for phospholipase A1, PLA1 and combines the necessary reagents to run 2 to 10 complete 96-well microplate assays that monitor activity in purified enzyme preparations and cell lysates. The importance of phospholipases in cellular signaling, lipid metabolism, inflammatory responses and pathological disorders related to these processes has stimulated demand for fluorescence-based activity monitoring methods. In particular the phospholipases resident in plasma and endothelium can perturb circulating LDL and HDL particles, creating pro-artherogenic forms. Recent evidence is drawing a link between these lipases and the progression of several severe neurodegenerative diseases, including Alzheimer's. PLA-1 improves upon two existing reagents in two ways: first it is now PLA-1 specific and secondly it has improved the assay quality by decreasing initial background noise.

Enzyme and Protein Activity Assays, Lipase and Phopholipase Activity, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage 1 kit. Store kit at -<20°C, protected from light, desiccated

Sufficient material is supplied for 200 reactions in 96 well microplates at 100 μl per well as described in the protocol below or 400 reaction in 384 low volume microplates at a volume of ≤ 25 μL per well.
Detection Method Fluorescence
For Use With (Application) Phospholipase Assay
Product Type EnzChek Assay Kit
Product Line EnzChek
Quantity 200 Reactions kit
Shipping Condition Room Temperature
Substrate Properties Lipid-Based Substrate, Chemical Substrate
Substrate Type Phospholipase Substrate
Target Enzyme Phospholipase
What is the LoD for the EnzChek Phospholipase A1 Assay Kit (Cat. No. E10219)?

The EnzChek Phospholipase A1 Assay Kit (Cat. No. E10219) can detect PLA1 at 0.04 U/mL or lower.

For Research Use Only. Not for use in diagnostic procedures.