missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EnzChek™ Paraoxonase Assay Kit
Description
For use with fluorescence intensity techniques
A highly sensitive, homogenous fluorometric assay for organophosphatase acitivty of paraoxonase. Based on hydrolysis of fluorogenic analog, this assay is >ten-fold more sensitive than the colorimetric assay, and unlike colorimetric assay, distinguishes samples of very similar paraoxonase activity.
- Assay requires only 5μL of serum, yields signal in as little as 15 minutes, and is linear for up to 50 minutes
- Assay requires only single reaction that can be either continuously monitored or terminated using the stop reagent included in kit
Cell Analysis, Cellular Imaging, Enzyme and Protein Activity Assays, High-Content Screening (HCS), Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Content And Storage | Store in freezer -5°C to -30°C. |
| Detection Method | Fluorescence |
| For Use With (Application) | Paraoxonase Assay |
| Product Type | EnzChek Assay Kit |
| Product Line | EnzChek |
| Quantity | 1 Kit |
| Shipping Condition | Room Temperature |
| Substrate Type | Paraoxonase Substrate |
| Target Enzyme | Paraoxonases |
For Research Use Only. Not for use in diagnostic procedures.