missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EnzChek™ Gelatinase/Collagenase Assay Kit, 250-2,000 assays

Catalog No. E12055
Encompass
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12055 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E12055 Supplier Invitrogen™ Supplier No. E12055

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fluorometric method for measuring gelatinase or collagenase activity

The EnzChek Gelatinase/Collagenase Assay Kit provides a sensitive, convenient and fast fluorometric method for measuring gelatinase or collagenase activity in purified enzyme systems, cell/tissue lysates or for screening inhibitors in a high-throughput format. The substrate in the EnzChek kit is fluorescein-labeled DQ gelatin conjugate that is highly labeled so that the fluorescence signal is quenched until enzymatic digestion yields highly fluorescent fragments. This substrate is also available separate from the kit (D-12054).

Cell Adhesion, Cell Analysis, Cell Viability, Proliferation and Function, Chemotaxis and Cell Migration, Enzyme and Protein Activity Assays, Protease/Peptidase Activity, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Gelatinase/Collagenase Assay
Product Type EnzChek Assay Kit
Product Line EnzChek
Quantity 1 Kit
Shipping Condition Room Temperature
Substrate Properties Protein-Based Substrate
Substrate Type Gelatinase/Collagenase Substrate
Target Enzyme Gelatinase/Collagenase

For Research Use Only. Not for use in diagnostic procedures.