missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EnzChek™ Elastase Assay Kit
Description
The EnzChek Elastase Assay Kit provides a sensitive, convenient and fast fluorometric method for measuring elastase or other protease activity in purified enzyme systems, cell/tissue lysates or for screening inhibitors in a high-throughput format. The substrate in the EnzChek kit is our BODIPY-FL-labeled DQ elastin conjugate that is highly labeled so that the fluorescence signal is quenched until enzymatic digestion yields highly fluorescent fragments.
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Content And Storage | Store in freezer -5°C to -30°C and protect from light. |
| Detection Method | Fluorescence |
| For Use With (Application) | Enzyme and Metabolic Activity |
| Product Type | EnzChek Assay Kit |
| Excitation Wavelength Range | 495 to 510 nm |
| Product Line | EnzChek |
| Quantity | 600 Assays |
| Shipping Condition | Room Temperature |
| Substrate Properties | Protein-Based Substrate |
| Substrate Type | Elastase Substrate |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.