missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EnzChek™ Direct Phospholipase C Assay Kit

Catalog No. E10215
Encompass
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10215 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E10215 Supplier Invitrogen™ Supplier No. E10215

Includes

1 kit

Monitors PC-PLC activity

This kit provides continuous monitoring of phosphotidyl choline specific phospholipase C (PLC) activity in microplate based biochemical assays. The assay uses a PCL selective fluorogenic substrate that emits a bright green emission upon cleavage. PC-PLC plays a crucial role in many cell signaling pathways involved in apoptosis and cell survival, as well as diseases ranging from cancer to HIV1-7. The assay uses a glycerophospho-ethanolamine with a dye-labeled sn-2 acyl chain as a substrate for PC-PLC. Substrate cleavage by PC-PLC before the phosphate releases the dye-labeled diacylglycerol, which produces a positive fluorescence signal that may be measured continuously. The reaction product has absorption and fluorescence emission maxima of 509nm and 516nm, respectively. Using purified enzyme from Bacillus cereus, the assay can detect as little as 10 mU/mL PC-PLC after one hour at room temperature. The kit has been proven useful for characterizing PC-PLC inhibition, and since it offers a direct measurement, the potential for false positives in a compound screen is eliminated.

  • Each kit provides enough reagents for 10 microplates, using 200μL volumes in 96 well format

Enzyme and Protein Activity Assays, Lipase and Phopholipase Activity, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room temperature

Specifications

Color Green
Content And Storage I kit Store kit at -<20°, protected from light, desiccated
Detection Method Fluorescence
For Use With (Application) Phospholipase Assay
For Use With (Equipment) Microplate Reader
Product Type EnzChek Assay Kit
Dye Type BODIPY™ FL
Emission 509/516
Filter Type FITC (Fluorescein) Filter
Product Line EnzChek
Quantity 1 Kit
Shipping Condition Room Temperature
Substrate Properties Lipid-Based Substrate, Chemical Substrate
Substrate Type Phospholipase Substrate
Target Enzyme Phosphatidylcholine-Specific Phospholipase C
Show More Show Less
What is the excitation/emission maxima of the reaction product when using the EnzChek Direct Phospholipase C Assay kit?

The fluorescence excitation/emission maxima is 509/516 nm.

For Research Use Only. Not for use in diagnostic procedures.