missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DiOC6(3) (3,3'-Dihexyloxacarbocyanine Iodide) DFS Item

For use with fluorescence microscopy

Supplier:  Invitrogen™ D273

Catalog No. D273


Only null left

Description

Description

DiOC6(3) is a cell-permeant, green-fluorescent, lipophilic dye that is selective for the mitochondria of live cells, when used at low concentrations. At higher concentrations, the dye may be used to stain other internal membranes, such as the endoplasmic reticulum.

Apoptosis, Cell Analysis, Cell Structure, Cell Viability, Proliferation & Function, Mitochondria, Mitochondria Function, Mitochondrial Function

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Green
Fluorescence
DiOC6(3)
Room Temperature
Store at room temperature and protect from light.
Fluorescence Microscope
100 mg
Cell Membranes & Lipids
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.