Learn More
Invitrogen™ DiO'; DiOC18(3) (3,3'-Dioctadecyloxacarbocyanine Perchlorate)
The green fluorescent, lipophilic carbocyanine >DiOC18(3) is widely used as a lipophilic tracer. It is weakly fluorescent in water, but highly fluorescent and quite photostable when incorporated into membranes.
Supplier: Invitrogen™ D275
Description
The green fluorescent, lipophilic carbocyanine DiOC18(3) is widely used as a lipophilic tracer. It is weakly fluorescent in water, but highly fluorescent and quite photostable when incorporated into membranes. It has an extremely high extinction coefficient and short excited-state lifetimes (∼1 nanosecond) in lipid environments. Once applied to cells, the dye diffuses laterally within the plasma membrane.
We recommend preparing stock solutions of lipophilic tracers in dimethyl formamide (DMF), dimethylsulfoxide (DMSO), or ethanol at 1 to 2.5 mg/mL. DMF is preferable to ethanol as a solvent for DiO.
Order Info
Shipping Condition: Room Temperature
Specifications
| Green | |
| 484 nm | |
| 100 mg | |
| 501 nm | |
| Liphophilic Tracer |
| Store at room temperature and protect from light. | |
| Fluorescence Microscope | |
| Fluorescence | |
| Room Temperature | |
| Cell Membranes, Lipids |
For Research Use Only. Not for use in diagnostic procedures.