missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Di-8-ANEPPS

Catalog No. D3167
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3167 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D3167 Supplier Invitrogen™ Supplier No. D3167
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Molecules that fluoresce in response to electrical potential changes in their environment

ANEP dyes are molecules that fluoresce in response to electrical potential changes in their environment. These are fast-response probes that operate by means of a change in their electronic structure, and consequently their fluorescence properties, in response to a change in the surrounding electric field. Their optical response is sufficiently fast to detect transient (millisecond) potential changes in excitable cells, including single neurons, cardiac cells, and intact brains. However, the magnitude of their potential-dependent fluorescence change is often small; fast-response probes typically show a 2-10% fluorescence change per 100 mV. Furthermore, these dyes display a potential-dependent shift in their excitation spectra, thus permitting the quantitation of membrane potential using excitation ratio measurements.

Potential-Sensitive ANEP Dye Specifications:

  • Zwitterionic molecule; is less susceptible to internalization than di-4-ANEPPS (Cat. No. D1199), permitting extended observation
  • Ex/Em maxima bound to model phospholipid membranes are ∼465/635 nm (but spectral properties are highly dependent on environment)
  • Nonfluorescent until bound to membranes
  • Soluble in ethanol, DMSO, and DMF (di-2-ANEPEQ (Cat. No. D6923)) is a water-soluble ANEP dye)
  • Dye is introduced into cells by direct addition of stock solution to cell culture medium, by using Pluronic™ F-127 (Cat. No. P6866), or by retrograde labeling
  • Fast-response probe, suitable for detecting submillisecond membrane potential changes

Applications for Potentiometric Probes

The plasma membrane of a cell typically has a transmembrane potential of approximately -70 mV (negative inside) as a consequence of K+, Na+, and Cl- concentration gradients that are maintained by active transport processes. Potentiometric probes offer an indirect method of detecting the translocation of these ions.

Increases and decreases in membrane potential-referred to as membrane hyperpolarization and depolarization, respectively-play a central role in many physiological processes, including nerve-impulse propagation, muscle contraction, cell signaling and ion-channel gating. Potentiometric probes are important tools for studying these processes.

Find More ANEP Dyes

We offer ANEP dyes in various forms. Review Fast-Response Probes-Section 22.2 in the Molecular Probes™ Handbook for more information on these probes.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.

Order Info

Shipping Condition: Room temperature

Specifications

Color Orange
Content And Storage Store at room temperature and protect from light.
Detection Method Fluorescence
For Use With (Application) Potential Changes
For Use With (Equipment) Fluorescence Microscope
Product Type ANEP Dye
Dye Type ANEP
Quantity 5 mg
Shipping Condition Room Temperature
Sub Cellular Localization Cell Membranes & Lipids
What are the excitation/emission maxima for Di-8-ANEPPS (Cat. No. D3167)?

Di-8-ANEPPS dye (Cat. No. D3167) is essent­ially nonfluorescent until bound to membranes. The excitation/emission maxima for Di-8-ANEPPS when bound to model phospholipid membranes are ~465/635 nm but spectral properties are highly dependent on the environment.

I am seeing high background outside of my neuronal cells when using membrane potential indicators. What can I do to reduce background?

If you use our FluoVolt Membrane Potential Kit (Cat. No. F10488), the kit provides a background suppressor to reduce this problem. For other indicators, consider the use of BackDrop Background Suppressor (Cat no. R37603, B10511, and B10512).

What is the difference between fast and slow-response membrane potential probes?

Molecules that change their structure in response to the surrounding electric field can function as fast-response probes for the detection of transient (millisecond) potential changes. Slow-response dyes function by entering depolarized cells and binding to proteins or membranes. Increased depolarization results in additional dye influx and an increase in fluorescence, while hyperpolarization is indicated by a decrease in fluorescence. Fast-response probes are commonly used to image electrical activity from intact heart tissues or measure membrane potential changes in response to pharmacological stimuli. Slow-responding probes are often used to explore mitochondrial function and cell viability.

What type of membrane potential indicators do you offer and how should I choose one for my experiment?

A membrane potential indicator selection guide can be found here (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cell-viability-and-regulation/ion-indicators/membrane-potential-indicators.html).

For Research Use Only. Not for use in diagnostic procedures.