missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DASPEI (2-(4-(dimethylamino)styryl) -N-Ethylpyridinium Iodide)

Catalog No. D426
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D426 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D426 Supplier Invitrogen™ Supplier No. D426

Stains mitochondria of live cells

DASPEI is a styryl dye that has a large fluorescence Stokes shift and is taken up relatively slowly as a function of membrane potential.

Cell Analysis, Cell Structure, Mitochondria, Mitochondrial Structure

Order Info

Shipping Condition: Room Temperature

Specifications

Color Yellow
Content And Storage Store at room temperature and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope
Product Type Dye
Quantity 1 g
Shipping Condition Room Temperature
Sub Cellular Localization Mitochondria

For Research Use Only. Not for use in diagnostic procedures.