missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ D-Luciferin, Potassium Salt

Catalog No. l2916
Encompass
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L2916 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L2916 Supplier Invitrogen™ Supplier No. L2916

Needed for luciferin/luciferase light-producing reaction

The ATP-dependent oxidation of luciferin by luciferase produces light, and this system is employed as a reporter in plants, bacteria, and mammalian cells. Because chemiluminescent techniques are virtually background-free, this reporter gene system is ideal for detecting low-level gene expression.

  • Exhibits physical properties identical to those of natural luciferin
  • Chemiluminescent after oxidation by luciferase
  • Luciferin potassium salt is readily soluble in aqueous buffers

Find More Fluorogenic and Photoactivatable Probes

A number of fluorogenic substrates, photoactivatable probes, and caged probes are available.

Cell Analysis, Enzyme and Protein Activity Assays, Luciferase Assays, Proteins, Expression, Isolation and Analysis, Reporter Assays

Order Info

Shipping Condition: Room temperature

Specifications

Type D-Luciferin
Quantity 25 mg
Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Chemiluminescence
Shipping Condition Room Temperature
Substrate Properties Chemical Substrate
Substrate Type Luciferase Substrate
Substrate Luciferin

For Research Use Only. Not for use in diagnostic procedures.