missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CoroNa™ Green, cell impermeant

Catalog No. C36675
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C36675 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C36675 Supplier Invitrogen™ Supplier No. C36675
Only null left

A sodium ion indicator that exhibits an increase in green fluorescence emission intensity upon binding Na+, with little shift in wavelength

  • Allows spatial and temporal resolution of Na+ concentrations in presence of physiological concentrations of other monovalent cations
  • Dye loads into cells more efficiently than does sodium green tetraacetate, and CoroNa Green indicator responds to broader range of Na+ concentration
  • With absorption/emission maxima of approximately 492/516nm for Na+-bound form, can be detected by any instrument that can detect fluorescein

Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling, Sodium and Potassium Detection

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope
Product Type Sodium Indicator
Dye Type Sodium Indicator
Product Line CoroNa
Quantity 1 mg
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.