Learn More
Invitrogen™ Click-iT™ RNA Alexa Fluor™ 594 Imaging Kit
Description
Includes
The kit contains sufficient material for 25, 18 x 18 coverslips.
The Click-iT RNA Alexa Fluor 594 Imaging Kit enables detection of global RNA transcription temporally and spatially in cells and tissues (PNAS (2008) 105:15779-84). The ability to detect newly synthesized RNA or changes in RNA levels resulting from disease, environmental damage or drug treatments is an important aspect of toxicological profiling. Utilizing an alkyne-modified nucleoside, 5-ethynyl uridine (EU), and powerful click chemistry, newly synthesized RNA can be detected without the use of radioactivity or antibodies with a simple, two-step procedure. In step one, the alkyne-containing nucleoside is fed to cells or animals and actively incorporated into nascent RNA. The small size of the tag enables efficient incorporation of the modified nucleoside into RNA, but not into DNA. Detection utilizes the chemoselective ligation or “click” reaction between and azide and an alkyne where the modified RNA is detected with a corresponding azide-containing dye. With its dimunitive “footprint”, the Click-iT detection molecule can easily penetrate complex samples and leaves open the possibility of multiplex analyses with other probes, including antibodies for the detection of RNA-interactive proteins for deeper biological insights.
- Less hazardous
Cell Analysis, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function, Cellular Imaging, Click Chemistries, DNA and RNA Purification and Analysis, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Staining and Detection, Labeling Chemistry, Nascent RNA Detection and Capture with Click Chemistry, Nucleic Acid Labeling and Oligo Synthesis, RNA Labeling
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Content And Storage | The kit contains sufficient material for 25, 18 X 18 coverslips. Store at 2-6°C, dessicate and protect from light. |
| Color | Red |
| Detection Method | Fluorescence |
| Format | Slide |
| Label or Dye | Alexa Fluor 594 |
| Excitation/Emission | 590/617 |
| For Use With (Equipment) | Confocal Microscope, Fluorescence Microscope |
| Product Line | Alexa Fluor, Click-iT, Molecular Probes |
| Product Type | New RNA Synthesis Assay |
| Quantity | 1 kit |
| Show More |
Frequently Asked Questions (FAQs)
Other than the EdU (Component A), DMSO (Component C), and Click-iT EdU buffer additive (Component F or G), all other components in the respective kits are not interchangeable. The types of reagents and amounts provided per kit were optimized either for imaging or flow cytometry. Using the imaging reagents may result in an excessive level of signal for flow cytometry detection and using the flow cytometry reagents may result in sub-optimal signal for imaging.
No. The cell must possess a pyrimidine salvage pathway; without this pathway, EU does not become phosphorylated to allow incorporation during RNA synthesis.
The fluorescence excitation/emission maxima is 590/615 nm.
For Research Use Only. Not for use in diagnostic procedures.