missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Click-IT™ ManNAz Metabolic Glycoprotein Labeling Reagent (tetraacetylated N-Azidoacetyl-D-Mannosamine)
GREENER_CHOICE

Catalog No. C33366
Encompass
Change view
Click to view available options
Quantity:
5.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C33366 5.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C33366 Supplier Invitrogen™ Supplier No. C33366

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Simple and robust two-step technique

The Click-iT ManNAz metabolic glycoprotein labeling reagent provides the first part of a simple and robust two-step technique to identify and characterize cell surface sialic acid-containing glycoproteins. In step one, cultured cells are incubated with the azide-modified mannosamine (ManNAz). The azido-sugar is metabolically incorporated into cell surface sialic acid-containing glycoproteins through the permissive nature of the oligosaccharide biosynthesis pathway. In step two, via the chemoselective ligation or click reaction between an azide and an alkyne, the azido-labeled glycoproteins can then be detected with a Click-iT Glycoprotein Detection kit for gels (TAMRA or Dapoxyl™ alkyne) or Western blots (biotin alkyne). These Click-iT products are compatible with LC-MS/MS and Multiplexed Proteomics™ technologies for in-depth analyses of the glycoproteome.

  • Less hazardous

Cell Analysis, Click Chemistries, Glycoprotein Analysis with Click Chemistry, Labeling Chemistry, Mass Spectrometry, Metabolic Labeling, Peptide Labeling, Protein and Antibody Labeling, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room temperature

Specifications

Product Type Labeling Reagent
Quantity 5.2 mg
Content And Storage Store in freezer -5°C to -30°C.
Detection Method Biotin-based, Fluorescence
Shipping Condition Room Temperature
Product Line Click-iT
Label or Dye Alexa Fluor 488, Alexa Fluor 555, Alexa Fluor 594, Alexa Fluor 647, Biotin, Oregon Green 488, TMR (Tetramethylrhodamine)
Labeling Target Proteins

For Research Use Only. Not for use in diagnostic procedures.