missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Click-IT™ Geranylgeranyl Alcohol, Azide, mixed isomers
GREENER_CHOICE

Catalog No. C10249
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10249 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C10249 Supplier Invitrogen™ Supplier No. C10249

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Achieve greater detection sensitivity in 1-D gels and western blots

Identify and characterize geranylated proteins with Click-iT geranylgeranyl alcohol, azide using the powerful click chemistry, a simple and robust two-step labeling and detection technique. In step one, the azide-containing biomolecule is fed to cells or animals and actively incorporated into proteins. Unlike other labels such as biotin or a fluorescent dye, the azide-tag is small enough that the tagged molecule is an acceptable substrate for the enzymes that incorporate this building block into proteins. Detection utilizes the chemoselective ligation or “click” reaction between and azide and an alkyne where the modified protein is detected with the corresponding alkyne-containing dye or hapten using either the Click-iT Cell Reaction Buffer Kit or the Click-iT Protein Buffer Kit. With the Click-iT Cell Reaction Buffer Kit, cells can be analyzed by fluorescence microscopy, flow cytometry or high-content imaging and analysis (HCS) together with other biomarkers of interest for content and context rich results. With the Click-iT Protein Reaction Buffer Kit, achieve detection sensitivity in 1-D gels and western blots in the low femtomole range or perform LC-MS/MS and MALDI MS analysis.

  • Less hazardous

Cell Analysis, Click Chemistries, Labeling Chemistry, Metabolic Labeling and Nascent Protein Synthesis with Click Chemistry

Order Info

Shipping Condition: Dry ice

Specifications

Product Type Geranylgeranyl Alcohol Azide
Quantity 1 mg
Content And Storage Store at ≤-20°C, desiccated and protected from light.
Labeling Method Metabolic Labeling
Shipping Condition Dry Ice
Product Line Click-iT, Molecular Probes
Label or Dye Azide
Format Solid
Labeling Target Proteins
I am observing no signal or very low signal for my click-labeled samples. What can I do to improve the signal?

The click reaction is only effective when copper is in the appropriate valency. Except for the DIBO alkyne-azide reaction, azides and alkynes will not react with each other without copper. Make sure that the click reaction mixture is used immediately after preparation when the copper (II) concentration is at its highest.
Do not use additive buffer that has turned yellow; it must be colorless to be active.
Cells need to be adequately fixed and permeabilized for the click reagents to have access to intracellular components that have incorporated the click substrate(s).
Some reagents can bind copper and reduce its effective concentration available to catalyze the click reaction. Do not include any metal chelator (e.g., EDTA, EGTA, citrate, etc.) in any buffer or reagent prior to the click reaction. Avoid buffers or reagents that include other metal ions that may be oxidized or reduced. It may be help to include extra wash steps on the cell or tissue sample before performing the click reaction.
You can repeat the click reaction with fresh reagents to try to improve signal. Increasing the click reaction time longer than 30 minutes will not improve a low signal. Performing a second, 30 minute incubation with fresh click reaction reagents is more effective at improving labeling.
Low signal can also be due to low incorporation of EdU, EU, or other click substrates. Other click substrates (e.g., AHA, HPG, palmitic acid, azide, etc.) incorporated into cellular components may have been lost if not adequately cross-linked in place or if the wrong fixative was used. For click substrates that are incorporated into the membrane or lipids, you should avoid the use of alcohol or acetone fixatives and permeabilizing agents.
The incorporated click substrate must be accessible at the time of the click reaction; labeling of incorporated amino acid analogs may be lower in native proteins relative to denatured proteins.
You may need to optimize the metabolic labeling conditions including analog incubation time or concentration. Cells that are healthy, not too high of a passage number and not too crowded may incorporate the analog better. You may create a positive control by including extra doses of the click substrate during multiple time points during an incubation time that spans or closely spans the doubling time of the cell type of interest.

For Research Use Only. Not for use in diagnostic procedures.