Learn More
Invitrogen™ Click-iT™ EdU Alexa Fluor™ 594 HCS Assay

Description
Includes
The kit contains sufficient material for two 96-well plates.
Mild fixation and detergent permeabilization sufficient for Click-iT EdU detection reagent to gain access to the DNA. The Click-iT EdU Alexa Fluor 594 HCS Assay is a superior alternative to traditional proliferation assays that is optimized for high-content imaging applications. In this assay the modified thymidine analogue EdU is efficiently incorporated into newly synthesized DNA and fluorescently labeled with a bright, photostable Alexa Fluor dye in a fast, highly specific click reaction. This fluorescent labeling of proliferating cells is accurate and compatible with antibody methods due to the mild click protocol. The most accurate proliferation-detection methods are based on the incorporation and measurement of nucleoside analogues in newly synthesized DNA, with bromodeoxyuridine (BrdU) a commonly used analogue. BrdU-labeled DNA is quantitated using anti-BrdU antibodies following DNA denaturation by harsh methods (HCl, heat, or enzymes) to expose the BrdU molecules. This step is time consuming and difficult to perform consistently. The harsh treatment can also adversely effect sample integrity and quality, which makes co-staining with other antibodies challenging.
- Simple: works the first time, every time, in less time
- Efficient: no denaturation steps or harsh treatment required
- Content-rich results: better preservation of cell morphology, antigen structure, and DNA integrity
- Consistent: not dependent on variable antibody lots for detection
- Less hazardous
Superior Proliferation Methodology
The Click-iT EdU Alexa Fluor 594 HCS Assay provides a superior alternative to BrdU assays for measuring cell proliferation. EdU (5-ethynyl-2'-deoxyuridine) is a nucleoside analog of thymidine and is incorporated into DNA during active DNA synthesis. With Click-iT EdU, mild fixation and detergent permeabilization is sufficient for the small molecule-based Click-iT EdU detection reagent to gain access to the DNA. As a consequence, the Click-iT EdU HCS assay is not only easy to use, but more accurate and compatible with cell cycle analysis and other intracellular or extracellular targets for truly content-rich results.
The Click-iTassay can be used on cells in culture or in vivo following administration of EdU by feeding or injection methods.
The Click-iTassay can be used with BrdU in dual pulse experiments by using the ant-BrdU (clone MoBu-1) antibody, which does not cross react with EdU.
The Click-iTtechnology is compatible with immunohistochemical, immunocytochemical, and fluorescent dyes that are fixation tolerant or designed for fixed-cell labeling.
Cell Analysis, Cell Cycle, Cell Proliferation, Cell Viability, Proliferation and Function, Cellular Imaging, DNA Replication and Repair, High-Content Screening (HCS)
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Color | Red |
| Content And Storage | The kit contains sufficient material for 2 96-well plates. Store at 2°C to 6°C, dessicate and protect from light. |
| Detection Method | Fluorescence |
| For Use With (Application) | High-content Screening (HCS) |
| For Use With (Equipment) | High Content Analysis Instrument |
| Product Type | HCS Assay |
| Dye Type | Alexa Fluor™ 594 |
| Format | 96-well plate |
| Product Line | Alexa Fluor, Click-iT, Molecular Probes |
| Quantity | 2 x 96 well plates |
| Show More |
Frequently Asked Questions (FAQs)
One may store the sample after fixation overnight in PBS at 4oC. For longer storage (<1 week) , store in buffer with 1-2% formaldehyde or in formalin to limit microbial growth. If you use sodium azide as a microbial inhibitor, it must be completely removed prior to the Click-iT reaction.
The problem is likely not the Alexa Fluor 594 azide. Since there are no alkynes endogenous to mouse tissue, there is nothing for the dye-azide to bind to. Since the background doesn't overlap with nuclei (DAPI signal), this isn't an issue of unintended EdU labeling. This red is autofluorescence from red blood cells; they autofluoresce in the red and don't have nuclei. This can be confirmed by checking a completely unlabeled tissue section (no dye present at all) to see if they are still present and by examining the cells at high magnification and looking for corpuscular shape.
Yes, but the standalone products are not shipped at the same amount as provided in the Click-iT EdU kits; the amount of dye-azide provided in the Click-iT kits is proprietary information. See these catalog numbers for the stand alone products:
- Cat. No. A10266: Alexa Fluor 488 azide
- Cat. No. A10270: Alexa Fluor 594 azide
- Cat. No. A10277: Alexa Fluor 647 azide
The click reaction is only effective when copper is in the appropriate valency. Azides and alkynes will not react with each other without copper. Make sure that the click reaction mixture is used immediately after preparation when the copper (II) concentration is at its highest.
Do not use additive buffer that has turned yellow; it must be colorless to be active.
Cells need to be adequately fixed and permeabilized for the TdT enzyme and click reagents to have access to the nucleus. Tissue samples require digestion with proteinase K or other proteolytic enzymes for sufficient TdT access.
Some reagents can bind copper and reduce its effective concentration available to catalyze the click reaction. Do not include any metal chelator (e.g., EDTA, EGTA, citrate, etc.) in any buffer or reagent prior to the click reaction. Avoid buffers or reagents that include other metal ions that may be o xidized or reduced. It may be help to include extra wash steps on the cell or tissue sample before performing the click reaction.
You can repeat the click reaction with fresh reagents to try to improve signal. Increasing the click reaction time longer than 30 minutes will not improve a low signal. Performing a second, 30 minute incubation with fresh click reaction reagents is more effective at improving labeling.
Your cells may not be apoptotic. Prepare a DNase I-treated positive control to verify that the TdT enzymatic reaction and click labeling reaction are working correctly.
The click reaction is very selective between an azide and alkyne. No other side reactions are possible in a biological system. Any non-specific background is due to non-covalent binding of the dye to various cellular components. The Select FX Signal Enhancer is not effective at reducing non-specific charge-based binding of dyes following the click reaction; we do not recommend its use with the Click-iT detection reagents. The best method to reduce background is to increase the number of BSA washes. You should always do a no-dye or no-click reaction control under the same processing and detection conditions to verify that the background is actually due to the dye and not autofluorescence. You should also perform the complete click reaction on a no-TdT enzyme control sample to verify the specificity of the click reaction signal.
For Research Use Only. Not for use in diagnostic procedures.