missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Click-iT™ AHA Alexa Fluor™ 488 Protein Synthesis HCS Assay
GREENER_CHOICE

Catalog No. c10289
Encompass
Change view
Click to view available options
Quantity:
2 plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10289 2 plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C10289 Supplier Invitrogen™ Supplier No. C10289

Includes

The kit contains sufficient reagents for performing two plates in a 96-well plate format.

Non-radioactive technique

The Click-iT AHA Alexa Fluor 488 Protein Synthesis HCS Assay provides a fast, sensitive, non-toxic and non-radioactive method for the detection of nascent protein synthesis utilizing fluorescence microscopy and high-content imaging. Click-iT AHA is an amino acid analog of methionine containing an azido moiety. Similar to 35S-methionine, Click-iT AHA is fed to cultured cells and incorporated into proteins during active protein synthesis. Detection of the incorporated amine acid utilizes a chemoselectiove ligation or 'click' reaction between an azide and an alkyne, where the azido modified protein is detected with the green-fluorescent Alexa Fluor 488 alkyne. The Click-iT AHA Alexa Fluor 488 Protein Synthesis HCS Assay has been successfully tested in A549 and U-2 OS cells with a variety of reagents that inhibit protein synthesis including cycloheximide, anisomycin and puromycin in both dose response and Min/Max format.

  • Less hazardous

Cell Analysis, Cell Metabolism, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function

Order Info

Shipping Condition: Room temperature

Specifications

Color Blue, Green
Content And Storage The kit contains sufficient reagents for performing 2 plates in a 96-well plate format.
  • Store at ≤-20°C, desiccate and protect from light.
  • Detection Method Fluorescence
    For Use With (Application) High-content Screening (HCS)
    For Use With (Equipment) Fluorescence Microscope, High Content Analysis Instrument
    Product Type HCS Assay
    Dye Type Alexa Fluor™ 488, Hoechst 33258
    Format 96-well plate
    Product Line Alexa Fluor, Click-iT, Molecular Probes
    Quantity 2 plates
    Shipping Condition Room Temperature
    Show More Show Less
    I am using Click-iT AHA (L-azidohomoalanine) kit to label nascent proteins in live cells, then detecting with TAMRA alkyne after fixation and permeabilization and a click reaction. But I'm seeing nucleolar labeling in the cells. It this expected?

    Yes. All proteins synthesized during the time the AHA is present will be detected, and they may be all over the cell. Our imaging shows strong labeling in the nucleoli and cytoplasm, as well as nuclear labeling.

    For Research Use Only. Not for use in diagnostic procedures.