missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CellLight™ Early Endosomes-GFP, BacMam 2.0

Catalog No. C10586
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10586 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C10586 Supplier Invitrogen™ Supplier No. C10586

Provides easy way to label early endosomes with green fluorescent protein in live cells

This ready-to-use construct is transfected into cells using BacMam 2.0 technology, where it expresses GFP fused to Rab5a. You can observe early endosomes-GFP behavior in live cells independently of organelle pH and label with multiple tracking or tracing dyes to image dynamic cellular processes.

Cells expressing CellLight constructs can also be fixed with formaldehyde for multiplexed imaging using immunocytochemical techniques.

BacMam Technology

CellLight Early Endosomes-GFP, BacMam 2.0, is a fusion construct of Rab5a and emGFP, providing accurate and specific targeting to cellular Early Endosomes-GFP. This fusion construct is packaged in the insect virus baculovirus, which does not replicate in human cells and is designated as safe to use with biosafety level (BSL) 1 in most laboratories. BacMam technology ensures that most mammalian cell types are transduced/transfected with high efficiency and minimal toxicity. This transient transfection can be detected after overnight incubation for up to five days—enough time to carry out most dynamic cellular analyses. Like any transfection/transduction technique, the BacMam method does not transfect/transduce all of the cells with equal efficiency, making it poorly suited to cellular population studies or automated imaging/counting. CellLight reagents are ideal for experiments where cellular or subcellular co-locatization is required, or for cellular function studies that need special resolution.

  • Fast and convenient: simply add CellLight reagent to cells, incubate overnight, and image—or store frozen, assay-ready cells for later use
  • Highly efficient: up to 90% transduction of a wide range of mammalian cell lines, including primary cells, stem cells, and neurons
  • Flexible: co-transduce more than one BacMam reagent for multiplex experiments or co-localization studies; tightly control expression levels by simply varying the dose
  • Less toxic: CellLight reagents are non-replicating in mammalian cells and are suitable for biosafety level (BSL) 1 handling
  • Flow Cytometer Laser Lines: 488

Cell Analysis, Cell Structure, Cell Tracing & Tracking, Cell Viability, Proliferation & Function, Cellular Imaging, Endocytosis, Exocytosis & Phagocytosis, Endosomes, Immunocytochemistry (ICC), Immunofluorescence (IF), Organelle Tracing

Order Info

Shipping Condition: Wet Ice

Specifications

Color Green
Content And Storage Store at 2°C to 6°C, protected from light. Do Not Freeze.
Excitation Wavelength Range 488⁄510
Dye Type GFP (EmGFP)
For Use With (Equipment) Confocal Microscope, Fluorescence Microscope
Quantity 1 mL
Detection Method Fluorescence
Emission Visible
Form Liquid
Product Line CellLight
Shipping Condition Wet Ice
Technique Fluorescence Intensity
Label Type Fluorescent Dye
Product Type Endosomes Label
Sub Cellular Localization Endosomes
Show More Show Less
How can I increase the transduction efficiency with the BacMam 2.0 reagents such as the the CellLight and Premo products?

Try varying particle-to-cell ratio (PPC), incubation volume, temperature and, cell density (if adherent cells are transduced). For adherent cells, we recommend a confluence of about 70%. Following the PPC, adjusting the volume is the next best parameter to change to optimize protein expression. If that doesn't work, you can also use the BacMam Enhancer Kit (Cat. No. B10107).

Is there any way to preserve the CellLights labeling beyond 5 days?

Cells transduced with the CellLights reagents can be stored frozen for several months after transduction, without loss of expression.

Are the CellLights products toxic to cells?

If the viral particles are used at the level we recommend, they are very well tolerated by cells.

For how long will the CellLights products label my cells?

The BacMam 2.0 CellLights typically express for 5 days after transduction.

What cell types can the CellLights products be used with?

The first generation BacMam reagents were shown to efficiently transduce over 90 cell types, including stable cell lines and primary cells. With BacMam 2.0, it is now possible to also efficiently transduce primary neurons and stem cells.

What safety precautions need to be taken to work with the BacMam reagent?

Our BacMam constructs do not replicate in mammalian cells, making them Biosafety Level I (the lowest biosafety level). They can be handled with the standard precautions used for any cell-based reagents.

Do CellLight BacMam 2.0 reagents work on all cell types?

BacMam delivery technology works with over 90 mammalian cell types. However, cells with hematopoietic origin do not show effective transduction with CellLight BacMam 2.0 reagents.

I keep getting low transduction efficiency when using CellLight labeling reagents on my neurons. What can I do to improve the efficiency?

Neurons are more difficult to transduce than many other cells. The main way to improve transduction is to label with a higher number of particles per cell. For primary neurons, it can also help to transduce them at the time of plating rather than on established cultures. There can also be a slower onset of expression in neurons and peak expression often occurs on day 2-3 rather than 16 hours after transduction.


For Research Use Only. Not for use in diagnostic procedures.