missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Calcium Green™-1 dextran, Potassium Salt, 10,000 MW, Anionic
Description
Labeled calcium indicators are molecules that exhibit an increase in fluorescence upon binding Ca2+
- Uses in many calcium signaling investigations, including measuring intracellular Ca2+, following Ca2+ influx and release, and multiphoton excitation imaging of Ca2+ in living tissues
- Cells may be physically loaded with the cell-impermeant salt forms of these dextran-conjugated indicators using patch pipet, microinjection, or Influx™ pinocyte-loading reagent
- Fluorescence signal from these cells is measured using fluorescence microscopy
- Dextran forms of calcium indicators show dramatic reduction in both leakage and compartmentalization compared to the AM ester forms
- Exhibit fluorescence increase upon binding Ca2+ with little shift in wavelength
- These probes are excited by visible light, and because the energy required for excitation is low, the potential for cellular photodamage is reduced
- Commonly used laser-based instruments (i.e., confocal laser scanning microscopes) are able to efficiently excite these indicators, and their emissions are in regions of the spectrum where cellular autofluorescence and scattering backgrounds are often less of a problem
- Label (Ex/Em): Calcium Green -1 (506/531nm)
- Fluorescence intensity increase upon binding Ca2+: approximately 14 fold
- Dextran size: 10,000 MW
Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Endocytosis, Exocytosis and Phagocytosis, Ionic Homeostasis and Signaling
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Content And Storage | Store in freezer -5°C to -30°C and protect from light. |
| Detection Method | Fluorescence |
| For Use With (Application) | Cell Viability and Proliferation |
| For Use With (Equipment) | Fluorescence Microscope |
| Product Type | Calcium Indicator |
| Dye Type | Fluorescent Dye-Based |
| Product Line | Calcium Green |
| Quantity | 5 mg |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.