missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Calcium Green™-1, AM, cell permeant

Catalog No. p-4922776
Encompass
Change view
Click to view available options
Quantity:
10 x 50 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C3011MP 500 μg
C3012 10 x 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C3011MP Supplier Invitrogen™ Supplier No. C3011MP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Labeled calcium indicators are molecules that exhibit an increase in fluorescence upon binding Ca2+

Labeled calcium indicators are molecules that exhibit an increase in fluorescence upon binding Ca2+. They have uses in many calcium signaling investigations, including measuring intracellular Ca2+, following Ca2+ influx and release, and multiphoton excitation imaging of Ca2+ in living tissues. Cells may be loaded with the AM ester forms of these calcium indicators by adding the dissolved indicator directly to dishes containing cultured cells. The fluorescence signal from these cells is generally measured using fluorescence microscopy, fluorescence microplate assays, or flow cytometry.

Calcium Indicator (AM Ester) Specifications:

  • Label (Ex/Em): Calcium Green™ -1 (506/531 nm)
  • Fluorescence intensity increase upon binding Ca2+ : ∼14 fold
  • Exhibit fluorescence increase upon binding Ca2+ with little shift in wavelength

Spectral Characteristics of Molecular Probes™ Calcium Indicators

These probes are excited by visible light, and because the energy required for excitation is low, the potential for cellular photodamage is reduced. Commonly used laser-based instruments (i.e., confocal laser scanning microscopes) are able to efficiently excite these indicators, and their emissions are in regions of the spectrum where cellular autofluorescence and scattering backgrounds are often less of a problem.

More Choices for Fluorescent Calcium Indicators

We offer a large selection of Molecular Probes™ calcium indicators for use in various experimental scenarios, for example dextran versions for reduced leakage and compartmentalization and BAPTA conjugates for detecting high-amplitude calcium transients. For more information, review Fluorescent Ca2+ Indicators Excited with Visible Light—Section 19.3 in the Molecular Probes™ Handbook.

For UV-excitable Ca2+ indicators, protein-based Ca2+ indicators, conjugates of Ca2+ indicators, and for fluorescence-based indicators of other metal ions (i.e., Mg2+, Zn2+) review Indicators for Ca2+, Mg2+, Zn2+ and Other Metal Ions—Chapter 19 in the Molecular Probes™ Handbook.

For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Calcium Indicator
For Use With (Equipment) Fluorescence Microscope
Product Type Calcium Indicator
Dye Type Calcium Green
Excitation Wavelength Range 495 to 510 nm
Product Line Calcium Green
Quantity 500 μg
Shipping Condition Room Temperature
Emission Wavelength Range 525 to 540 nm
Excitation Wavelength 506 nm
Emission Wavelength 531 nm
Show More Show Less
I am doing calcium flux imaging with your Fura-2 calibration kit, but am seeing a large variability in ratio in different places around the slide. I am correcting for uniform illumination, using the product as directed, and sealing the coverslip with nail polish.

The nail polish may be the problem. The Kd value (calcium sensitivity) changes depending upon the dye's environment. Nail polish has solvents that can leech under the coverslip and cause variability. We recommend either going without a sealing or sealing with melted paraffin painted on the coverslip edges with a cotton-tipped applicator (paraffin is hydrophobic and has no solvents).

I need to label cells with Fluo-4, AM, for a calcium flux assay. How long after labeling will the dye be retained?

After loading dye into the cells, intracellular esterases remove the 'AM' moiety from the dye. When the 'AM' group is removed, the dye is able to bind calcium and fluoresce. Since the dye is not covalently bound to any cellular components, it may be actively effluxed from the cell. The rate of efflux is dependent upon the inherent properties of the cell, culture conditions and other factors. The dye may be retained for hours, days or even weeks or lost in a matter of minutes. The use of Probenecid (Cat. No. P36400) limits loss by active efflux.

For Research Use Only. Not for use in diagnostic procedures.