missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CellTrace™ BODIPY™ TR Methyl Ester (Lipophilic Counterstain For GFP)

Catalog No. C34556
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C34556 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C34556 Supplier Invitrogen™ Supplier No. C34556

Retained after paraformaldehyde fixation

  • Readily permeates cell membranes and selectively stains mitochondria and endomembranous organelles such as endoplasmic reticulum and Golgi apparatus, but does not appear to localize in plasma membrane

Cell Adhesion, Cell Analysis, Cell Proliferation, Cell Tracing and Tracking, Cell Viability, Proliferation and Function, Cell-Based Reporter Assays, Cellular Imaging, Cellular Toxicology Assays, Chemotaxis and Cell Migration, Drug Discovery and Development, Enzyme and Protein Activity Assays, Fluorescent Protein (e.g. GFP) Assays, Fluorescent Protein Assays (GFP), General Cell Tracing, High-Content Screening (HCS), Immunofluorescence (IF), Immunofluorescence Staining and Detection, Proteins, Expression, Isolation and Analysis, Reporter Assays, Target-Based ADME/Tox Assays

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Dye Type BODIPY Dyes
Quantity 1 mL
Product Line BODIPY, CellTrace
Reagent Type Cell Tracker Compounds, Cell Labeling Reagents
Shipping Condition Room Temperature
Label Type Other Labels or Dyes
Product Type Lipophilic Tracer
I am storing the DMSO stock solutions of the CellTrace cell proliferation reagents and am not obtaining a good signal with my cells compared to freshly dissolved dye stock. Why does the product not work after storage?

We provide the CellTrace reagents in small aliquots and strongly recommend discarding any unused DMSO/dye stocks. The CellTrace reagents have acetyl groups to cap the charges on the dyes to make them cell permeant, and succinimidyl ester amine-reactive moiety to allow for covalent attachment to cellular components for long-term retention. Both acetyl groups and succinimidyl esters will readily hydrolyze if any water is present during storage. DMSO is hygroscopic and thus readily absorbs water from the atmosphere. If you must store your dye stocks, you will need to use a good quality, anhydrous DMSO stock that has not been opened often and store the vial within an air-tight container containing some desiccant to keep the DMSO/dye stock solution anhydrous during storage.

I am using the CellTrace Cell Proliferation reagents and am not obtaining good separation of my cell generation peaks. How can I improve the peak separation?

Here are some tips to obtain uniform staining and a bright, unstimulated parent generation peak:

Dissolve the CellTrace dye stock immediately before use in the DMSO provided in the kit or in good quality, anhydrous DMSO to obtain the best reactivity and cell permeability.
Stain in PBS or other amine-free, protein-free physiological buffer. Do not stain in medium.
Start with a single-cell suspension and gently agitate the cells during staining.
Quickly remove the unbound dye by incubating the cells in ice-cold media for 5 minutes and then wash twice more with pre-warmed media.
Include a dead-cell stain in the assay and gate only on live cells.
Analyze as many cells as possible from each sample.
Use a low flow rate for analysis on hydrodynamic focusing cytometers.
A good staining concentration for the CellTrace dyes is generally within 1-10 µM, but the optimal concentration for a particular cell type will vary. Observe your cells in a stain dilution series to determine the optimal concentration for your cells.
Some cell types may take up dye with a broad staining intensity distribution. If this is the case for your cells, then you will need to do an initial sort of the stained, unstimulated parent cells to select for a narrow peak distribution.

Can I store the stock solutions of the CellTrace reagents and how do you recommend storing them?

We provide the CellTrace reagents in small aliquots and strongly recommend discarding any unused DMSO dye stocks. Some of the CellTrace reagents have diacetate groups to cap the charges on the dyes to make them cell permeant, and some have succinimidyl ester amine-reactive groups for long-term cellular retention. Both diacetates and succinimidyl esters will readily hydrolyze if any water is present during storage. DMSO is very hygroscopic and thus readily absorbs water from the atmosphere. We do not recommend storing stock solutions of CellTrace reagents because storage of the product in solution will inevitably lead to partial or complete loss of reactivity.

How do the CellTrace Cell Proliferation reagents work?

CellTrace Cell Proliferation reagents are all cell-permeant dyes that are cleaved by intracellular esterases to yield highly fluorescent compounds that also covalently bind to cellular amines, attaching the dye to various cellular components and providing a very stable signal. These reagents show little cytotoxicity with minimal observed effects on the proliferative ability of many cells.

I am trying to assay cell proliferation with a CellTrace stain, and I am not seeing separate peaks for each cell division. How can I optimize this assay?

The key to good generational profiles with CellTrace reagents is starting with cells that are evenly labeled so that they have a tight coefficient of variance (CV) when run at time zero after labeling. If the peak is too broad, the generations will overlap each other and the series of peaks will become a hump. Even labeling can be achieved by starting with a uniform cell population (not a mixture of lymphocytes and granulocytes for example) as staining will be proportional to cell size. Cells are labeled rapidly, so you want to pre-dilute the dye and mix it into your cells rapidly. Be sure that the cells are not sitting in a clump in the bottom of your tube. The easiest way to do this is to make a 2x dye solution (1x = 1-10 µM) and resuspend your cells in a half volume of medium (no serum or BSA). Add the dye to the cells and invert a few times to mix. Gently agitate the cells during staining. Once the dye incubation is over (20 min, 37 degrees C), add serum or BSA (at least 1%) to scavenge any remaining unreacted dye. Spin down cells, wash 1x, and resuspend in complete medium. After a 10-20 min incubation to undergo de-esterification, cells are ready to be set up for whatever treatment you are planning. Be sure to keep a time zero control as you need to know where the first generation ran.

What are the advantages of flow cytometry?

-Measures data from single cells.
-Data are obtained for a large number of cells, generating a rich statistical analysis of cell populations.
-Because single cells are measured, it will reveal heterogeneity within a population.
-With the ability to multiplex, small sub-populations can be identified.
-Thousands of cells can be analyzed rapidly.
-It is ideally suited for blood samples and other cells in suspension.
-Data can be re-analyzed multiple times after acquisition.
-Flow cytometry files (FCS) can be archived.

What kinds of applications can I run on a flow cytometer?

There are several applications, some of which include immunophenotyping, cell cycle analysis, apoptosis assays such as annexin V staining, CellEvent Caspase-3/7 assay, and TUNEL assay, cell viability, proliferation assays such as CellTrace assay and Click-iT EdU assay, measurements of mitochondrial potential with MitoProbe assays, and cell counting using counting beads.


For Research Use Only. Not for use in diagnostic procedures.