missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BCEF, AM (2',7'-Bis-(2-Carboxyethyl)-5-(and-6)-Carboxyfluorescein, Acetoxymethyl Ester) Solution
Description
- BCECF, AM is cell-permeant, dual-excitation ratiometric pH indicator with a pKa of approximately 6.98
- Intracellular pH measurements with BCECF are made by determining pH-dependent ratio of emission intensity (detected at 535nm) when dye is excited at approximately 490nm versus emission intensity when excited at its isobestic point of approximately 440nm
Cell Adhesion, Cell Analysis, Cell Metabolism, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function, Cell-Based Ion Channel Assays, Drug Discovery and Development, Ion Channel Biology, Ionic Homeostasis and Signaling, Target and Lead Identification and Validation, pH Detection
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Concentration | 1 mg⁄ml |
| Content And Storage | Store in freezer -5°C to -30°C. |
| Detection Method | Fluorescence |
| For Use With (Equipment) | Fluorescence Microscope, Flow Cytometer, Microplate Reader |
| Product Type | BCEF AM pH indicator |
| Dye Type | pH Indicator |
| Quantity | 1 mL |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.