missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BCECF Acid (2',7'-Bis-(2-Carboxyethyl)-5-(and-6)-Carboxyfluorescein), mixed isomers

Catalog No. b1151
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1151 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B1151 Supplier Invitrogen™ Supplier No. B1151

Dual-excitation ratiometric pH indicator

  • For measuring changes ± 1 pH unit around pH 7
  • pKa of approximately 6.98
  • pH measurements with BCECF are made by determining pH-dependent ratio of emission intensity (detected at 535nm) when dye is excited at approximately 490nm versus emission intensity when excited at its isobestic point of approximately 440nm

Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling, pH Detection

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store at room temperature and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Microplate Reader
Product Type pH indicator
Dye Type pH Indicator
Quantity 1 mg
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.