missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BAPTA, Tetrapotassium Salt, cell impermeant

Catalog No. B1204
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1204 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B1204 Supplier Invitrogen™ Supplier No. B1204

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Highly selective chelator for Ca2+ over mg2+ and can be used to control level of extracellular Ca2+

  • BAPTA is more selective for Ca2+ than EDTA and EGTA, and its metal binding is also less sensitive to pH

Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store at room temperature.
Product Type BAPTA Tetrapotassium Salt
Dye Type Chelator (Non-Fluorescent)
Quantity 1 g
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.