Learn More
Invitrogen™ Amplex™ Red Uric Acid/Uricase Assay Kit
Description
Detects uric acid or uricase using either a fluorescence microplate reader or fluorometer
The Amplex™ Red Uric Acid/Uricase Assay Kit provides a sensitive and simple method for detecting uric acid or uricase using either a fluorescence microplate reader or fluorometer.
- Detect concentrations as low as 100 nM uric acid or 0.2 mU/mL of uricase activity
- Format allows for multiple time point measurements
- Designed for minimal autofluorescence interference
Serum uric acid is the end product of purine metabolism in the body tissues and is cleared through the kidneys by glomerular filtration. Most animals can metabolize uric acid to more readily excreted products, but humans lack the necessary enzyme, urate oxidase (uricase), as a result of the presence of two 'nonsense mutations' in the human gene for uricase.
The Amplex™ Red Uric Acid/Uricase Assay Kit provides an ultrasensitive method for detecting uric acid or for monitoring uricase activity. In the assay, uricase catalyzes the conversion of uric acid to allantoin, hydrogen peroxide, and carbon dioxide. In the presence of horseradish peroxidase (HRP), hydrogen peroxide reacts stoichiometrically with the Amplex™ Red reagent to generate the red-fluorescent oxidation product resorufin.
Because resorufin has absorption and fluorescence emission maxima of approximately 571 nm and 585 nm, respectively, there is little interference from autofluorescence in most biological samples.
Use Amplex™ Red Assays for a Broad Range of Investigations
A wide variety of validated Amplex™ Red assays are available for studying cell signaling and lipids, neurobiology, inflammation and immune function, and metabolism. We also offer Amplex™ UltraRed Reagent (Cat. No. A36006), a second-generation reagent providing greater sensitivity and brighter fluorescence, and the Amplex™ Red/UltraRed Stop Reagent (Cat. No. A33855). The Amplex™ Red/UltraRed Stop Reagent provides convenience and control by allowing the fluorescence signal-generating reaction to be terminated at a user-determined time point. After addition of the stop reagent, the fluorescence signal remains stable for at least three hours. Custom assay design and packaging are also available.
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Content And Storage | Store in freezer -5°C to -30°C and protect from light. |
| Detection Method | Fluorescence |
| For Use With (Application) | Enzyme and Metabolic Activity |
| For Use With (Equipment) | Microplate Reader, Spectrophotometer, Fluorometer |
| Product Type | Amplex Red Assay Kit |
| Dye Type | Amplex Red |
| Excitation Wavelength Range | 570 to 585 nm |
| Format | 96-well Plate |
| Product Line | Amplex |
| Quantity | 400 Assays |
| Show More |
Frequently Asked Questions (FAQs)
The components of Krebs-Ringer buffer (salts) should not cause oxidation of the Amplex reagent (which, in the presence of peroxidase and H2O2 oxidizes to resorufin, which is pink in color and fluorescent). Try water alone (the water used to make the Krebs-Ringer buffer). Since Hank's Buffered Saline Solution is typically purchased rather than made in the lab, it likely would not have the same contaminant. Another option is to degas the buffer prior to use to removed dissolved oxygen radicals.
This is not recommended. The presence of endogenous proteases can complicate the assay by degrading the horseradish peroxidase (HRP). Endogenous peroxidases and antioxidants can modify the H2O2 required for the reaction, competing with HRP (and catalase) for the substrate.
The Amplex Red Assays are best performed with either purified enzymes or extracted H2O2 in a defined buffer system, extracellular solutions or body fluids (media, serum, etc.) that do not exhibit high levels of endogenous protease or oxidase activity and do not contain antioxidants.
For Research Use Only. Not for use in diagnostic procedures.