Learn More
Invitrogen™ Amplex™ Red/UltraRed Stop Reagent
Description
Designed for use with both the Amplex Red and Amplex UltraRed fluorogenic substrates and assay kits
Amplex Red/UltraRed Stop Reagent provides convenience and control by allowing the fluorescence signal-generating reaction to be terminated at a user-determined time point. enic substrates and assay kits. It is designed to terminate reactions containing up to 0.1 units/mL of horseradish peroxidase (HRP) and 5μM hyrogen peroxide. After addition of the stop reagent, the fluorescence signal remains stable for at least 3 hours.
- Sensitive biomolecular assays based on hydrogen peroxide–generating enzyme systems linked to peroxidase–mediated oxidation of the fluorogenic Amplex UltraRed or Amplex Red substrates
- Detection reactions are performed in microplate wells and are initiated by adding fluorogenic Amplex UltraRed or Amplex Red substrate, resulting in a continuous fluorescence increase that proceeds for 30 minutes or more
- Unknown analyte concentrations are determined by comparing fluorescence intensities measured at a certain time point during the reaction to parallel measurements at same time point on standard samples of known concentration
- It is critical to ensure that timing of standard and unknown sample measurements is same
Build Your Own Immunoassay, Cell Analysis, Enzyme and Protein Activity Assays, Immunoassays, Lipase and Phopholipase Activity, Oxidase and Peroxidase Activity, Proteins, Expression, Isolation and Analysis, Ready-To-Use Immunoassay
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Content And Storage | Store in refrigerator at 2°C to 8°C |
| For Use With (Application) | Protein Assays |
| Product Type | Stop Solutions |
| Form | Lyophilized |
| Product Line | Amplex |
| Quantity | 1 set |
| Shipping Condition | Room Temperature |
Frequently Asked Questions (FAQs)
This is not recommended. The presence of endogenous proteases can complicate the assay by degrading the horseradish peroxidase (HRP). Endogenous peroxidases and antioxidants can modify the H2O2 required for the reaction, competing with HRP (and catalase) for the substrate.
The Amplex Red Assays are best performed with either purified enzymes or extracted H2O2 in a defined buffer system, extracellular solutions or body fluids (media, serum, etc.) that do not exhibit high levels of endogenous protease or oxidase activity and do not contain antioxidants.
For Research Use Only. Not for use in diagnostic procedures.