missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Amplex™ Red and Amplex™ UltraRed Reagents

Catalog No. A12222
Encompass
Change view
Click to view available options
Quantity:
5 mg
5 x 1 mg
10 x 10 mg
Dye Type:
Amplex Red Reagent
Amplex UltraRed Reagent
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Dye Type
A12222 5 mg Amplex Red Reagent
A22177 10 x 10 mg Amplex Red Reagent
A36006 5 x 1 mg Amplex UltraRed Reagent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A12222 Supplier Invitrogen™ Supplier No. A12222

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Amplex Red and UltraRed reagents are fluorogenic substrates for peroxidase. These highly sensitive and stable probes react with H2O2 in the presence of HRP for solution-based assays and ELISAs. Amplex UltraRed is a brighter version of Amplex Red with an improved pH range and sensitivity.

The Amplex Red and Amplex UltraRed reagents are highly sensitive and stable fluorescent probes for the detection of hydrogen peroxide (H2O2) and ideal fluorogenic substrates for peroxidase. These reagents can be used for solution-based assays to detect the activity of H2O2 released from biological samples or with HRP-based ELISA applications. The Amplex UltraRed reagent is an improved version of Amplex Red that is brighter and more sensitive to peroxidase with less pH sensitivity compared to the Amplex Red reagent.

In the presence of horseradish peroxidase (HRP), the Amplex Red and UltraRed reagents react in a 1:1 stoichiometry with H2O2 to produce highly fluorescent, strongly absorbing resorufin (ex/em ∼570/585 nm) or Amplex UltraRed (ex/em ∼568/581 nm), respectively. These oxidized reaction products have a broad red emission with little interference from blue and green autofluorescence found in most biological samples. Because H2O2 is produced in many different enzymatic reactions, the Amplex Red and UltraRed reagents coupled with HRP can be used to detect the activity of many different enzymes. These reagents are also used to detect H2O2 released from biological samples, including cells and isolated mitochondria.

Features of the Amplex Red reagent:

  • Sensitive and fast detection of HRP activity or H2O2 for solution-based assays and ELISAs
  • Detection of H2O2 produced in many different enzymatic reactions
  • Measurement with either fluorescence or absorbance, permitting either fluorogenic or chromogenic detection

The Amplex Red reagent has been used to develop a variety of novel fluorogenic and chromogenic assays for enzymes that produce hydrogen peroxide. When coupled with added horseradish peroxidase, these assays that generate H2O2 permit the ultrasensitive quantitation of a diverse assortment of analytes, including glucose, galactose, cholesterol, glutamic acid, xanthine (or hypoxanthine), uric acid, choline, and acetylcholine.

The Amplex UltraRed reagent offers brighter fluorescence and enhanced sensitivity on a per-mole basis in peroxidase or peroxidase-coupled enzyme assays compared to Amplex Red. The Amplex UltraRed reagent is also less sensitive to acidic pH and exhibits improved stability in the presence of H2O2 or thiols such as dithiothreitol (DTT). For HRP-based ELISA applications, this results in at least two-fold greater sensitivity compared to Amplex Red.

Benefits of the Amplex UltraRed reagent over Amplex Red:

  • Brighter signals and enhanced sensitivity for HRP activity and H2O2
  • Less sensitive to acidic pH
  • Improved stability in the presence of H2O2 or thiols such as DTT

The Amplex Red and Amplex UltraRed reaction products resorufin and Amplex UltroxRed have high extinction coefficients so they can be measured either fluorometrically or spectrophotometrically with absorbance at ∼560 nm, providing flexibility in detection of H2O2, various enzyme assays, and HRP-based ELISAs.

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage Store in freezer (-5°C to -30°C).
Detection Method Fluorescence
For Use With (Application) Hydrogen Peroxidase Assay
For Use With (Equipment) Fluorometer, Microplate Reader
Product Type Red Hydrogen Peroxide
Dye Type Amplex Red Reagent
Form Solid
Product Line Amplex
Quantity 5 mg
Shipping Condition Room Temperature
Substrate HRP (Horseradish Peroxidase) Substrate
Substrate Properties Chemical substrate
Validated Application Cell Analysis
Show More Show Less
I'm using an Amplex Red kit, the reagent changes color to pink almost immediately in my own Krebs-Ringer buffer but not in HBSS. Why is this?

The components of Krebs-Ringer buffer (salts) should not cause oxidation of the Amplex reagent (which, in the presence of peroxidase and H2O2 oxidizes to resorufin, which is pink in color and fluorescent). Try water alone (the water used to make the Krebs-Ringer buffer). Since Hank's Buffered Saline Solution is typically purchased rather than made in the lab, it likely would not have the same contaminant. Another option is to degas the buffer prior to use to removed dissolved oxygen radicals.

Is it possible to use the Amplex Red Reagent (Cat. No. A12222) with a colorimetric microplate reader and measure the optical density (OD)? What would be the ideal wavelength for OD measurements?

Yes it can be used for a colorimetric assay. In general, absorbance is less sensitive than fluorescence so the sensitivity of the colorimetric assay may be lower. You may need to empirically determine the detection limit and linear dynamic range for the colorimetric assay. Resorufin, the fluorescent product of Amplex Red, has an absorbance peak at 570 nm.

I have ordered and tested your Amplex Red Reagent (Cat. Nos. A12222, A22177), but it is not reacting correctly. What could be the cause?

Either the horseradish peroxidase or other enzymes in the assay may not be active, the H2O2 may have degraded, or the sample could have some component that is consuming H2O2 or inactivating the enzymes required for the assay. We recommend that you check when the reagent was purchased and how it was stored prior to use; the reagent should not be used past the warranty period.

Do you recommend using Amplex Red Reagent (Cat. Nos. A12222, A22177) in live cells?

Amplex Red is cell impermeant, but also its conversion to resorufin requires direct contact with the reaction of peroxidase with H2O2 (in solution, at the surface, or within cells). Within cells, peroxidases and H2O2 may not be sufficiently localized or active to provide enough signal.

Can I use Amplex Red Reagent (Cat. Nos. A12222, A22177) in the presence of catalases?

Unless you are specifically analyzing catalase activity, as with the Amplex Red Catalase Assay Kit (Cat. No. A22180), the assay should be devoid of catalases. Catalases compete with the horseradish peroxidase for H2O2; the conversion of Amplex Red to resorufin requires the reaction of horseradish peroxidase with H2O2.

How should I store Amplex Red Reagent (Cat. Nos. A12222, A22177)?

We recommend storing Amplex Red Reagent under vacuum (anaerobic conditions) and dehydrated. Do not store Amplex Red Reagent in solution.

What is the color of Amplex Red Reagent (Cat. Nos. A12222, A22177)?

Amplex Red Reagent is white/off-white or slightly pink (due to minor spontaneous oxidation during storage/shipping).

Can Amplex Red Assays be performed using cell lysates?

This is not recommended. The presence of endogenous proteases can complicate the assay by degrading the horseradish peroxidase (HRP). Endogenous peroxidases and antioxidants can modify the H2O2 required for the reaction, competing with HRP (and catalase) for the substrate.

The Amplex Red Assays are best performed with either purified enzymes or extracted H2O2 in a defined buffer system, extracellular solutions or body fluids (media, serum, etc.) that do not exhibit high levels of endogenous protease or oxidase activity and do not contain antioxidants.


For Research Use Only. Not for use in diagnostic procedures.