missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Amplex™ Red Glutamic Acid/Glutamate Oxidase Assay Kit

Catalog No. A12221
Encompass
Change view
Click to view available options
Quantity:
200 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12221 200 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A12221 Supplier Invitrogen™ Supplier No. A12221

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Simple method for detecting glutamic acid/glutamate oxidase activity.

The Amplex Red Glutamic Acid/Glutamate Oxidase Assay Kit provides a sensitive and simple method for detecting glutamic acid/glutamate oxidase activity using a fluorescence microplate reader or fluorometer. The Amplex Red Glutamic Acid/Glutamate Oxidase Assay Kit provides an ultrasensitive method for detecting glutamic acid or for continuously monitoring glutamate oxidase activity using a fluorescence microplate reader or fluorometer. In the assay, L-glutamic acid is oxidized by glutamate oxidase to produce α-ketoglutarate, NH3, and hydrogen peroxide. L-alanine and L-glutamate–pyruvate transaminase are included in the reaction to regenerate L-glutamic acid by transamination of α-ketoglutarate, resulting in multiple cycles of the initial reaction and a significant amplification of the hydrogen peroxide produced. The hydrogen peroxide reacts with 10-acetyl-3,7-dihydroxyphenoxazine (Amplex Red reagent) in a 1:1 stoichiometric ratio in a reaction catalyzed by horseradish peroxidase (HRP) to generate the highly fluorescent product resorufin. Because resorufin has absorption and fluorescence emission maxima of approximately 571nm and 585nm, respectively, there is little interference from autofluorescence in most biological samples.

  • Detect concentrations as low as 10nm L-glutamic acid or 40μU/mL of purified L-glutamate oxidase
  • Format allows for multiple time point measurements
  • Designed for minimal autofluorescence interference
  • Custom assay design and packaging are also available

Use Amplex Red Assays for a Broad Range of Investigations

A wide variety of validated Amplex Red assays are available for studying cell signaling and lipids, neurobiology, inflammation and immune function, and metabolism. We also offer Amplex UltraRed Reagent, a second-generation reagent providing greater sensitivity and brighter fluorescence, and the Amplex Red/UltraRed Stop Reagent. The Amplex Red/UltraRed Stop Reagent provides convenience and control by allowing the fluorescence signal-generating reaction to be terminated at a user-determined time point. After addition of the stop reagent, the fluorescence signal remains stable for at least three hours.

Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Enzyme and Protein Activity Assays, Oxidase and Peroxidase Activity, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Glutamic Acid/Glutamate Oxidase Assay
For Use With (Equipment) Fluorometer, Microplate Reader
Product Type Amplex Red Assay Kit
Dye Type Other Label(s) or Dye(s)
Format 96-well plate
Product Line Amplex
Quantity 200 Assays
Shipping Condition Room Temperature
I'm using an Amplex Red kit, the reagent changes color to pink almost immediately in my own Krebs-Ringer buffer but not in HBSS. Why is this?

The components of Krebs-Ringer buffer (salts) should not cause oxidation of the Amplex reagent (which, in the presence of peroxidase and H2O2 oxidizes to resorufin, which is pink in color and fluorescent). Try water alone (the water used to make the Krebs-Ringer buffer). Since Hank's Buffered Saline Solution is typically purchased rather than made in the lab, it likely would not have the same contaminant. Another option is to degas the buffer prior to use to removed dissolved oxygen radicals.

Can Amplex Red Assays be performed using cell lysates?

This is not recommended. The presence of endogenous proteases can complicate the assay by degrading the horseradish peroxidase (HRP). Endogenous peroxidases and antioxidants can modify the H2O2 required for the reaction, competing with HRP (and catalase) for the substrate.

The Amplex Red Assays are best performed with either purified enzymes or extracted H2O2 in a defined buffer system, extracellular solutions or body fluids (media, serum, etc.) that do not exhibit high levels of endogenous protease or oxidase activity and do not contain antioxidants.

For Research Use Only. Not for use in diagnostic procedures.