missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Amplex™ Red Galactose/Galactose Oxidase Assay Kit

Catalog No. A22179
Encompass
Change view
Click to view available options
Quantity:
400 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22179 400 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A22179 Supplier Invitrogen™ Supplier No. A22179
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Detects galactose or monitors galactose oxidase activity.

The Amplex™ Red Galactose/Galactose Oxidase Assay Kit provides a sensitive and simple method for detecting galactose oxidase activity or galactose using a fluorescence microplate reader or fluorometer.

  • Detect concentrations as low as 2 mU/mL of galactose oxidase activity or 4 μM galactose
  • Format allows for multiple time point measurements
  • Designed for minimal autofluorescence interference

Galactose oxidase is an important tool in the detection of galactose in biological fluids. The Amplex™ Red Galactose/Galactose Oxidase Assay Kit provides an ultrasensitive method for detecting galactose or for monitoring galactose oxidase activity. Galactose oxidase catalyzes the oxidation of galactose at the C6 position and generates hydrogen peroxide. In the presence of horseradish peroxidase (HRP), hydrogen peroxide reacts in a 1:1 stoichiometric ratio with the Amplex™ Red reagent to generate the red-fluorescent compound resorufin.

Because resorufin has absorption and fluorescence emission maxima of approximately 571 nm and 585 nm, respectively, there is little interference from autofluorescence in most biological samples.

Use Amplex™ Red Assays for a Broad Range of Investigations

A wide variety of validated Amplex™ Red assays are available for studying cell signaling and lipids, neurobiology, inflammation and immune function, and metabolism. We also offer Amplex™ UltraRed Reagent (Cat. No. A36006), a second-generation reagent providing greater sensitivity and brighter fluorescence, and the Amplex™ Red/UltraRed Stop Reagent (Cat. No. A33855). The Amplex™ Red/UltraRed Stop Reagent provides convenience and control by allowing the fluorescence signal-generating reaction to be terminated at a user-determined time point. After addition of the stop reagent, the fluorescence signal remains stable for at least three hours. Custom assay design and packaging are also available.

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Enzyme and Metabolic Activity
For Use With (Equipment) Microplate Reader, Spectrophotometer, Fluorometer
Product Type Amplex Red Assay Kit
Dye Type Amplex Red
Excitation Wavelength Range 570 to 585 nm
Format 96-well Plate
Product Line Amplex
Quantity 400 Assays
Shipping Condition Room Temperature
Emission Wavelength Range 570 to 585 nm, 585 to 600 nm
Excitation Wavelength 571 nm
Emission Wavelength 585 nm
Show More Show Less
I'm using an Amplex Red kit, the reagent changes color to pink almost immediately in my own Krebs-Ringer buffer but not in HBSS. Why is this?

The components of Krebs-Ringer buffer (salts) should not cause oxidation of the Amplex reagent (which, in the presence of peroxidase and H2O2 oxidizes to resorufin, which is pink in color and fluorescent). Try water alone (the water used to make the Krebs-Ringer buffer). Since Hank's Buffered Saline Solution is typically purchased rather than made in the lab, it likely would not have the same contaminant. Another option is to degas the buffer prior to use to removed dissolved oxygen radicals.

Can Amplex Red Assays be performed using cell lysates?

This is not recommended. The presence of endogenous proteases can complicate the assay by degrading the horseradish peroxidase (HRP). Endogenous peroxidases and antioxidants can modify the H2O2 required for the reaction, competing with HRP (and catalase) for the substrate.

The Amplex Red Assays are best performed with either purified enzymes or extracted H2O2 in a defined buffer system, extracellular solutions or body fluids (media, serum, etc.) that do not exhibit high levels of endogenous protease or oxidase activity and do not contain antioxidants.

For Research Use Only. Not for use in diagnostic procedures.