missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Vybrant™ Alexa Fluor™ 555 Lipid Raft Labeling Kit
Description
Vybrant Lipid Raft Labeling Kits include cholera toxin subunit B (CT-B) labeled with either the green-fluorescent Alexa Fluor 488, the orange-fluorescent Alexa Fluor 555 or the red-fluorescent Alexa Fluor 594 dye, as well as an antibody that specifically recognizes CT-B. A 10X solution of phosphate-buffered saline (10X PBS), pH 7.2, and detailed instructions are also included in the kits.
Cell Analysis, Cell Structure, Cell Tracing & Tracking, Lipid Rafts, Organelle Tracing
Order Info
Shipping Condition: Wet Ice
Specifications
Specifications
| Content And Storage | Store in freezer -5°C to -30°C and protect from light. |
| Quantity | 1 kit |
| Product Line | Alexa Fluor |
| Protein Subtype | Cholera Toxin |
| Shipping Condition | Wet Ice |
| Label Type | Alexa Fluor Dyes |
| Product Type | Lipid Raft Labeling Kit |
| Sub Cellular Localization | Cell Membranes and Lipids |
For Research Use Only. Not for use in diagnostic procedures.