missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Vybrant™ Alexa Fluor™ 488 Lipid Raft Labeling Kit

Catalog No. V34403
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V34403 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V34403 Supplier Invitrogen™ Supplier No. V34403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Provides convenient, reliable and extremely bright fluorescent labeling of lipid rafts in live cells

Vybrant Lipid Raft Labeling Kits include cholera toxin subunit B (CT-B) labeled with either the green-fluorescent Alexa Fluor 488, the orange-fluorescent Alexa Fluor 555 or the red-fluorescent Alexa Fluor 594 dye, as well as an antibody that specifically recognizes CT-B. A 10X solution of phosphate-buffered saline (10X PBS), pH 7.2, and detailed instructions are also included in the kits.

Cell Analysis, Cell Structure, Cell Tracing & Tracking, Lipid Rafts, Organelle Tracing

Order Info

Shipping Condition: Wet Ice

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Quantity 1 kit
Product Line Alexa Fluor
Protein Subtype Cholera Toxin
Shipping Condition Wet Ice
Label Type Alexa Fluor Dyes
Product Type Lipid Raft Labeling Kit
Sub Cellular Localization Cell Membranes and Lipids

For Research Use Only. Not for use in diagnostic procedures.