missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Isolectin GS-IB4 From Griffonia simplicifolia, Alexa Fluor™ 647 Conjugate

Catalog No. I32450
Encompass
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I32450 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I32450 Supplier Invitrogen™ Supplier No. I32450

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Isolated from the seeds of Griffonia simplicifolia, a tropical African legume

Isolectin GS-IB4 is a 114,000-dalton glycoprotein that is part of a family of five tetrameric type I isolectins (IA4, IA3B, IA2B2, IAB3, and IB4). The A subunit prefers N-acetyl-D-galactosamine end groups while the B subunit is selective for terminal a-D-galactosyl residues. The far-red fluorescent Alexa Fluor 647 isolectin GS-IB4 conjugate can be used in fluorescence microscopy or flow cytometry.

Cell Analysis, Cell Tracing & Tracking, Cellular Imaging, Flow Cytometry, Flow Cytometry Antibodies & Secondary Detection, General Cell Tracing, Immunofluorescence (IF), Immunofluorescence Staining & Detection, In Vivo Imaging, Microbial Tracking

Order Info

Shipping Condition: Room Temperature

Specifications

Quantity 500 μg
Source Africa Shrub legume
Conjugate Alexa Fluor 647
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label Type Alexa Fluor Dyes
Shipping Condition Room Temperature
Form Lyophilized
Protein Subtype Lectins
Product Line Alexa Fluor

For Research Use Only. Not for use in diagnostic procedures.