missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Acridine Orange (10 mg/mL in water)

Catalog No. A3568
Encompass
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A3568 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A3568 Supplier Invitrogen™ Supplier No. A3568

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cell-permeant nucleic acid binding dye

Acridine Orange emits green fluorescence when bound to dsDNA and red fluorescence when bound to ssDNA or RNA. This unique characteristic makes acridine orange useful for cell-cycle studies. Acridine orange has also been used as a lysosomal dye. This dye is available in solution (A-3568).

Apoptosis, Cell Analysis, Cell Cycle, Cell Structure, Cell Tracing & Tracking, Cell Viability & Cytotoxicity, Cell Viability, Proliferation & Function, DNA Fragmentation, Nucleus, Nucleoli & Nuclear Envelope, Organelle Tracing

Specifications

Color Orange
Content And Storage Store in refrigerator 2°C to 8°C and protect from light.
Excitation Wavelength Range 460/650 (RNA)
Dye Type Cell-Permeant
For Use With (Equipment) Fluorescence Microscope
Quantity 10 mL
Detection Method Fluorescence
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Dye
Sub Cellular Localization Nucleic Acids, Nucleus
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.