missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ACMA (9-Amino-6-Chloro-2-Methoxyacridine)

Catalog No. a1324
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1324 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1324 Supplier Invitrogen™ Supplier No. A1324

DNA intercalator that selective binds to poly (d(A-T))

Excitation of the ACMA-DNA complex (excitation/emission maxima ∽419/483nm) is possible with most UV-light sources, making it compatible for use with both shorter- and longer-wavelength dyes. ACMA also apparently binds to membranes in the energized state and becomes quenched if a pH gradient forms. It has been extensively employed to follow cation and anion movement across membranes and to study the proton-pumping activity of various membrane-bound ATPases.

Cell Analysis, Cell Structure, Cell Viability, Proliferation & Function, Cellular Imaging, Immunofluorescence (IF), Ionic Homeostasis & Signaling, Nucleus, Nucleoli & Nuclear Envelope, pH Detection

Order Info

Shipping Condition: Room Temperature

Specifications

Color Blue
Content And Storage Store at room temperature and protect from light.
Excitation Wavelength Range 419⁄483
For Use With (Equipment) Fluorescence Microscope
Quantity 100 mg
Detection Method Fluorescence
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Nucleic Acid Stain
Sub Cellular Localization Cell Membranes, Lipids, Nucleus

For Research Use Only. Not for use in diagnostic procedures.