missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 5-CFDA, AM (5-Carboxyfluorescein Diacetate, Acetoxymethyl Ester)

Catalog No. c1354
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1354 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C1354 Supplier Invitrogen™ Supplier No. C1354

A cell-permeant esterase substrate that can serve as a viability probe

  • Measures both enzymatic activity, which is require to activate its fluorescence, and cell-membrane integrity, which is required for intracellular retention of their fluorescent product
  • Electronically neutral AM ester of CFDA can be loaded into cells at lower concentrations than CFDA
  • Upon hydrolysis by intracellular esterases, this AM ester also yields carboxyfluorescein: As compared with fluorescein, carboxyfluorescein contains extra negative charges and is therefore better retained in cells

Cell Adhesion, Cell Analysis, Cell Counting and Viability, Cell Counting, Viability, and Cryopreservation, Cell Culture, Cell Proliferation, Cell Tracing and Tracking, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function, Cellular Imaging, Cellular Toxicology Assays, Chemotaxis and Cell Migration, Drug Discovery and Development, General Cell Tracing, High-Content Screening (HCS), Immunofluorescence (IF), Immunofluorescence Staining and Detection, Ionic Homeostasis and Signaling, Target-Based ADME/Tox Assays, pH Detection

Order Info

Shipping Condition: Room temperature

Specifications

Content And Storage Store in freezer -5°C to -30°C.
Product Type 5-CFDA AM
Dye Type Other Label(s) or Dye(s)
Quantity 5 mg
Reagent Type CFDA & Related Compounds
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.