missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 2-NBDG (2-(N-(7-Nitrobenz-2-oxa-1,3-diazol-4-yl)Amino)-2-Deoxyglucose)

Catalog No. N13195
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N13195 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N13195 Supplier Invitrogen™ Supplier No. N13195

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

2-NBDG is a fluorescent glucose analog that has been used to monitor glucose uptake in live cells, as an indicator of cell viability.

2-NBDG is a fluorescent glucose analog that has been used to monitor glucose uptake in live cells, as an indicator of cell viability. Although sensitive to its environment NBD fluorescence typically displays excitation/emission maxima of ∼465/540 nm and can be visualized using optical filters designed for fluorescein.

Dissolve the dye in water or 1X PBS (pH 7.2) to prepare a stock solution up to 10 mg/mL. Stock solutions should be prepared fresh and used on same day.

Order Info

Shipping Condition: Room Temperature

Specifications

Content And Storage Store in freezer (-5°C to -30°C) and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
Product Type 2-NBDG
Dye Type Other Label(s) or Dye(s)
Format Tube(s), Slide(s)
Quantity 5 mg
Shipping Condition Room Temperature
What are the final concentrations of 2-NBDG and 6-NBDG, and incubation times that may be used?

Final concentrations of 2-NBDG can range from 10 µM to 600 µM; 100 µM to 600 µM have been used with bacteria and yeasts and, from 10 µM to 200 µM for primary and cultured mammalian cells.

Final concentrations of 6-NBDG may range from 30 µM up to 300 µM.

Cells should be incubated at the desired temperature and time to allow for sufficient detection of green-yellow fluorescence within cells. Uptake is temperature dependent. Incubation times may range from seconds up to 30 minutes or longer, dependent upon the final concentration of the reagent, cell type, culture conditions, and other factors.

What solvents can be used to resolubilize either 2-NBDG or 6-NBDG to make a stock solution, and how should this stock solution be stored?

Compatible solvents (and approximate maximum solubility): DMSO ( ˜10 mg/mL), DMF (˜10 mg/mL), ethanol (˜20 mg/mL), methanol (˜20 mg/mL), H2O (˜10 mg/mL), PBS, pH 7.2 (˜10 mg/mL).

Stock solutions may be prepared using the recommended solvents in concentrations at or below their maximum solubility and then stored frozen, desiccated (for non-aqueous solvents), and protected from light. Aqueous solutions should not be stored longer than one day.

For long-term storage, these reagents should be stored as a solid at ≤–20°C. To make smaller aliquots, dissolve the reagent in ethanol, make smaller aliquots in separate vials and then evaporate the solvent using a vacuum pump. Do not use DMSO or DMF due to their low vapor pressure.

For Research Use Only. Not for use in diagnostic procedures.