missing translation for 'onlineSavingsMsg'
Learn More

Molecular Grade Water, Cytiva Hyclone™

Catalog No. WFIMGW20L
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
WFIMGW20L 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. WFIMGW20L Supplier Cytiva Supplier No. WFIMGW20L
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Molecular Grade Water is quality tested and certified RNase, DNase, and protease free.

  • Every lot contains zero traces of toxic agents such as DEPC
  • Well suited for use in diverse molecular biology applications
  • Manufactured under cGMP and ISO 13485 guidelines in a FDA registered facility
  • Purified utilizing a nine-step process resulting in the highest quality water on the market
  • Distilled
  • Deionized

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Color Clear, Colorless
Grade Molecular Certified
Packaging Cubitainer
Certificate of Analysis Yes
Quantity 20 L
ShelfLife 36 Months
Recommended Storage 2-30°C