Learn More
Thermo Scientific™ Molecular BioProducts Conical-Bottom Centrifuge Tubes
Description
- 1.5 mL conical bottom tube
- Natural color, nonsterile
- Bagged tube and unattached cap with O-ring
- Tubes made of virgin PP
- O-ring made of EPDM
Specifications
Specifications
| Capacity (Metric) | 1.5 mL |
| Color | Natural |
| Material | Plastic |
| Packaging | Bag |
| Sterility | Non-sterile |
| Bottom Shape | Conical |
| Max. RCF | 20,000 x g |