Learn More
Moldex™ HandyStrap™ Series R95 Particulate Respirators
R95 disposable respirators feature a unique HandyStrap™ with buckle, allowing the mask to hang down around the user’s neck when not in use.
Supplier:  Moldex™ 2940R95
 View more versions of this product
Catalog No. 19-999-367
Only null left
Explore available promotions
Description
Description
  • Cloth strap is cool and comfortable against the skin
  • HandyStrap™ with buckle, allows mask to hang down around the user’s neck when not in use preventing premature disposal
  • Dura-Mesh™ provides bi-directional protection from dirt on the outside and moisture on the inside
  • Soft foam nose cushion for added comfort and no pressure points
  • Softspun™ lining for increased comfort and durability
  • Ventex™ exhalation valve lets the hot air out faster to enable easier breathing
  • Meets heat and flame resistance in accordance with ANSI/ISEA 110-2003 Section 7.11.1
  • R95 Series Applications: Grinding, torch cutting, sanding, metal pouring, sweeping, metal working, bagging, foundries, stone quarrying, agriculture, metal machining, construction, cement, ceramics manufacturing, underground mining, timber/lumber mills, polishing, buffing, lime, poultry, textiles, welding, brazing, soldering
  • R95 Plus Nuisance Levels of Ozone and Organic Vapor Applications (Fisher cat #19-046-756 and 19-046-757 only): Welding, arc cutting, foundry operations, brazing, plasma spraying, laser beam cutting/welding, laboratory settings, agriculture, petrochemical manufacturing, rendering plants, auto repair, mining operations, waste disposal
  • R95 Plus Nuisance Levels of Acid Gases Applications (Fisher cat #19-999-367 and 19-999-368 only): Welding, arc cutting, foundry operations, brazing, plasma spraying, laser beam cutting/welding, laboratory settings, agriculture, petrochemical manufacturing, rendering plants, auto repair, mining operations, waste disposal
Specifications
Specifications
R - Somewhat Resistant to Oil
NIOSH 42 CFR 84, R95
Cup/Molded
R95
Particulate Respirator with Nuisance Acid Gas Relief
Yes
Use when oil aerosols are not present, including paper mills, aluminum reduction, coal-fired utilities and petrochemicals
2940R95
Acid Gases
HandyStrap™ with Buckle
ANSI/ISEA 110-2003, Section 7.11.1
Gray
Single-use
Dura-Mesh
Medium/Large
95%
Cloth
Soft foam
10/Pk.
No
Product Suggestions
Product Suggestions