Learn More
Moldex™ HandyStrap™ Series R95 Particulate Respirators
R95 disposable respirators feature a unique HandyStrap™ with buckle, allowing the mask to hang down around the user’s neck when not in use.
Supplier:  Moldex™ 2840R95
 View more versions of this product
Catalog No. 19-046-756
Only null left
Explore available promotions
Description
Description
  • Cloth strap is cool and comfortable against the skin
  • HandyStrap™ with buckle, allows mask to hang down around the user’s neck when not in use preventing premature disposal
  • Dura-Mesh™ provides bi-directional protection from dirt on the outside and moisture on the inside
  • Soft foam nose cushion for added comfort and no pressure points
  • Softspun™ lining for increased comfort and durability
  • Ventex™ exhalation valve lets the hot air out faster to enable easier breathing
  • Meets heat and flame resistance in accordance with ANSI/ISEA 110-2003 Section 7.11.1
  • R95 Series Applications: Grinding, torch cutting, sanding, metal pouring, sweeping, metal working, bagging, foundries, stone quarrying, agriculture, metal machining, construction, cement, ceramics manufacturing, underground mining, timber/lumber mills, polishing, buffing, lime, poultry, textiles, welding, brazing, soldering
  • R95 Plus Nuisance Levels of Ozone and Organic Vapor Applications (Fisher cat #19-046-756 and 19-046-757 only): Welding, arc cutting, foundry operations, brazing, plasma spraying, laser beam cutting/welding, laboratory settings, agriculture, petrochemical manufacturing, rendering plants, auto repair, mining operations, waste disposal
  • R95 Plus Nuisance Levels of Acid Gases Applications (Fisher cat #19-999-367 and 19-999-368 only): Welding, arc cutting, foundry operations, brazing, plasma spraying, laser beam cutting/welding, laboratory settings, agriculture, petrochemical manufacturing, rendering plants, auto repair, mining operations, waste disposal
Specifications
Specifications
R - Somewhat Resistant to Oil
NIOSH 42 CFR 84, R95
Cup/Molded
R95
Particulate Respirator with Nuisance Organic Vapor Relief
Yes
Welding, soldering and brazing, grinding, sanding, sweeping, bagging, stone quarrying, metal machining, cement, underground mining, polishing, buffing, poultry, agriculture, chemical processing, printing, food processing, paint and varnish manufacturing,
2840R95
Ozone and Organic Vapors
HandyStrap™ with Buckle
ANSI/ISEA 110-2003, Section 7.11.1
Gray
Single-use
Dura-Mesh
Medium/Large
95%
Cloth
Soft foam
10/Pk.
No
Product Suggestions
Product Suggestions