Learn More
Moldex™ Glide™ Foam No Roll Twist-In Ear Plugs
Glide™ Trio’s unique multi-curved stem rotates for a custom fit inside each ear canal. Simply push and twist to angle desired fit.
Supplier:  Moldex™ MDX6940
 View more versions of this product
Catalog No. 19384356
Only null left
Explore available promotions
Description
Description
  • Unique multi-curved stem rotates for a custom fit
  • Large handle on curved stem for easy insertion and removal
  • No roll down design is perfect for workers with dirty hands or wearing gloves
  • Cord can be attached at any desired length
  • Large soft foam pod for a comfortable, snug seal
Specifications
Specifications
Yes
Universal
No
100 Pairs/Pk.
Green
Uncorded
Reusable
Class AL
Construction, Utilities, Manufacturing, Production, Mining, Oil and Gas, and Other Various Industries
Poly bag
Foam
30 dB
Bell
No
Product Suggestions
Product Suggestions