missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MOG Recombinant Rabbit Monoclonal Antibody (JM23-06)
GREENER_CHOICE

Catalog No. PIMA532857
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532857 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532857 Supplier Invitrogen™ Supplier No. MA532857
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

The product of this gene is a membrane protein expressed on the oligodendrocyte cell surface and the outermost surface of myelin sheaths. Due to this localization, it is a primary target antigen involved in immune-mediated demyelination. This protein may be involved in completion and maintenance of the myelin sheath and in cell-cell communication. Alternatively spliced transcript variants encoding different isoforms have been identified.

Specifications

Antigen MOG
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JM23-06
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene MOG
Gene Accession No. Q16653, Q61885, Q63345
Gene Alias B230317G11Rik; BTN6; BTNL11; DAQB-92E24.2; dystonia 2, torsion (autosomal recessive); MGC26137; MOG; MOG alpha 6; MOG alpha-5; MOG alpha-6; MOG AluA; MOG AluB; MOG Ig AluB; MOG Ig-AluB; MOGIG2; MOGIG-2; myelin oligodendrocyte glycoprotein; myelin-oligodendrocyte glycoprotein; NRCLP7
Gene Symbols MOG
Host Species Rabbit
Immunogen Synthetic peptide within Human Myelin oligodendrocyte glycoprotein aa 51-100.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17441, 24558, 4340
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less