missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MMP8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA5149489
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA5149489 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA5149489 Supplier Invitrogen™ Supplier No. PA5149489

Goat Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

The MMP8 gene encodes matrix metalloproteinase-8, also known as collagenase-2, an enzyme belonging to the matrix metalloproteinase (MMP) family, which plays a pivotal role in the breakdown of extracellular matrix (ECM) components. MMP8 is primarily involved in the degradation of type I and III collagen, facilitating tissue remodeling and repair processes. This enzyme is produced by neutrophils and is active in areas of tissue inflammation and injury, where it modulates the ECM to aid in wound healing. MMP8 is implicated in various physiological processes, including embryonic development, angiogenesis, and bone remodeling. However, excessive or unregulated activity of MMP8 can lead to pathological conditions such as arthritis, periodontal disease, and cancer metastasis, where ECM degradation contributes to disease progression. Research into MMP8 aims to understand its regulation and involvement in disease states, providing insights into potential therapeutic strategies for targeting MMP activity in inflammatory and degenerative conditions.

Specifications

Antigen MMP8
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with trehalose and no preservative
Gene MMP8
Gene Accession No. P22894
Gene Alias BB138268; CLG1; Collagenase; Collagenase 2; collagenase-2; HNC; matrix metallo protease; matrix metallopeptidase 8; matrix metallopeptidase 8 (neutrophil collagenase); matrix metalloproteinase 8; matrix metalloproteinase 8 (neutrophil collagenase); Matrix metalloproteinase8; matrix metalloproteinase-8; MMP; MMP 8; Mmp8; MMP-8; MMPs; Neutrophil collagenase; neutrophil collagenease; PMN leukocyte collagenase; PMNL CL; PMNL collagenase; PMNLCL; PMNL-CL
Gene Symbols MMP8
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human MMP-8, Phe21-Gly467.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4317
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less